Thermo Scientific™

Random Access Storage Option for the Orbitor™ RS2 Microplate Mover for QuantStudio™ Real-time PCR Systems

Catalog number: A43782
Have Questions?
Thermo Scientific™

Random Access Storage Option for the Orbitor™ RS2 Microplate Mover for QuantStudio™ Real-time PCR Systems

Catalog number: A43782
Have Questions?
Catalog Number
A43782
Unit Size
1 each
Price (USD)
Catalog NumberUnit SizePrice (USD)
A437821 eachRequest A Quote
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
This Random Access Storage Option is designed for use with the Orbitor RS2 Microplate Mover and a QuantStudio real-time PCR system, enabling real-time PCR in automated environments for increased sample throughput.

This storage option holds up to 24 plates/3 hotels (384-well). The hotels enable users to easily add new plates or change the order in which the plates are run. Hotels are ideal when there are multiple users using 384-well plates. This option is not ideal for 96-well plates.

Couple this Random Access Storage Option with the Orbitor RS2 Microplate Mover for QuantStudio Systems to complete your automation bundle.

For integration of two QuantStudio systems with the Orbitor RS2 mover, the Dual QuantStudio System Integration Kit is required in addition to the automation bundle.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment)
QuantStudio 7 Pro, QuantStudio 7 Flex, or QuantStudio 12 K Flex Real-Time PCR Systems and The Orbitor RS2 Microplate Mover
For Use With (Application)
Real Time PCR (qPCR)
Shipping Condition
Room Temperature
Description
Storage Option
Quantity
1 each

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text