SuperSignal™ Western Blot Substrate Bundle, Pico PLUS + trial-size Femto
SuperSignal™ Western Blot Substrate Bundle, Pico PLUS + trial-size Femto
Thermo Scientific™

SuperSignal™ Western Blot Substrate Bundle, Pico PLUS + trial-size Femto

This substrate kit is comprised of SuperSignal West Pico PLUS Chemiluminescent Substrate plus a sample size of SuperSignal West FemtoRead more
Have Questions?
Catalog NumberQuantity
A43840500 mL bundle
Catalog number A43840
Price (USD)
404.65
Online Exclusive
448.00
Save 43.35 (10%)
Each
Add to cart
Quantity:
500 mL bundle
Price (USD)
404.65
Online Exclusive
448.00
Save 43.35 (10%)
Each
Add to cart
This substrate kit is comprised of SuperSignal West Pico PLUS Chemiluminescent Substrate plus a sample size of SuperSignal West Femto Maximum Sensitivity Substrate to provide chemiluminescent detection substrates for both routine and high sensitivity western blotting.

SuperSignal West Pico PLUS Chemiluminescent Substrate
Thermo Scientific SuperSignal West Pico PLUS Chemiluminescent Substrate enables picogram to high femtogram-level protein detection by western blot analysis. It is best for routine, everyday western blotting applications, providing excellent signal intensity, sensitivity and duration across a wide range of targets.

Sensitivity: pictogram to femtogram sensitivity
Stability: 8-hr working solution stability; 1-year kit stability at room temperature
Compatibility: nitrocellulose and PVDF membranes
Signal duration: 6 to 24 hours
Recommended primary antibody concentration: 1:1,000–1:5,000 dilution (0.2–1.0 μg/mL)
Recommended secondary antibody concentration: 1:20,000–100,000 dilution (10–50 ng/mL)

View full product information ›

SuperSignal West Femto Maximum Sensitivity Substrate
Thermo Scientific SuperSignal West Femto Maximum Sensitivity Substrate is an ultra-sensitive enhanced chemiluminescent (ECL) substrate for low femtogram protein level detection by western blot analysis with horseradish peroxidase (HRP) enzyme. It is optimized to detect low abundant and/or difficult to detect targets.

Sensitivity: low-femtogram (mid-zeptomole)
Stability: 8-hr working solution stability, 1-year kit stability at 4°C
Compatibility: nitrocellulose and PVDF membranes
Signal duration: up to 8 hours
Recommended primary antibody concentration: 1:5,000–1:100,000 dilution (10–200 ng/mL)
Recommended secondary antibody concentration: 1:100,000–1:500,000 dilution (2–10 ng/mL)

View full product information ›

For Research Use Only. Not for use in diagnostic procedures.
Specifications
SensitivityPicogram to femtogram, low femtogram (mid-zeptomole)
Shipping ConditionRoom Temperature
Sufficient For60 Mini-Gel Blots, 5000 sq. cm of membrane; 2 Mini-Gel Blots, 200 sq. cm of Membrane
Detection MethodChemiluminescence
For Use With (Application)Western Blot
FormLiquid
Membrane CompatibilityNitrocellulose, PVDF
Product LineSuperSignal™
Product TypeSubstrate
Quantity500 mL bundle
SubstrateHorseradish Peroxidase
Unit SizeEach
Contents & Storage
• 500 mL SuperSignal West Pico PLUS reagent, store at room temperature
• 20 mL SuperSignal West Femto reagent, store at 2–8°C

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link