Pierce™ Endotoxin-Free Water
Pierce™ Endotoxin-Free Water
Thermo Scientific™

Pierce™ Endotoxin-Free Water

Pierce Endotoxin-Free Water is free of endotoxins and enzymes, including proteases. Using Endotoxin-Free Water helps eliminate the risk of harmfulRead more
Promo ImagePromotion
Promo code:RPUZZ25
Stock up on essentials to piece your discovery together
Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzleLearn More
Have Questions?
Catalog NumberQuantity
A43883Promo Image4 x 50 mL
Catalog number A43883
Price (USD)
20.69
Each
In stock
Add to cart
Quantity:
4 x 50 mL
Price (USD)
20.69
Each
Add to cart
Pierce Endotoxin-Free Water is free of endotoxins and enzymes, including proteases.
Using Endotoxin-Free Water helps eliminate the risk of harmful activity produced by bacteria that can interfere with cell health and function. It is suitable for usage in molecular biology or general biochemistry applications and contains <0.005 EU/mL endotoxin.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionEndotoxin-Free Water
Endotoxin<0.005 EU/mL
PackagingBottle(s)
Purification MethodSterile filtered with 0.5 μm filter
SterilitySterile
Quantity4 x 50 mL
Unit SizeEach

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection