Pierce Plant Total Protein Extraction Kit
Pierce Plant Total Protein Extraction Kit
Thermo Scientific™

Pierce Plant Total Protein Extraction Kit

The Thermo Scientific Pierce Plant Total Protein Extraction Kit effectively extracts protein from all kinds of dry and fresh plantRead more
Catalog NumberQuantity
A4405650 preps
Catalog number A44056
Price (JPY)
36,300
Online offer
Ends: 26-Sep-2025
57,200
Save 20,900 (37%)
Each
Contact Us ›
Quantity:
50 preps
Ask our AI about this Product
The Thermo Scientific Pierce Plant Total Protein Extraction Kit effectively extracts protein from all kinds of dry and fresh plant tissue without liquid nitrogen or organic solvents.

Features of the Plant Total Protein Extraction Kit:
Efficient—obtain excellent protein yields in less than 10 minutes using a single buffer and a spin column
Versatile—native or denaturing lysis buffers for protein extraction from leaves, stems, and seeds
Compatible—extracts can be quantified using the BCA Protein Assay Kit or the Rapid Gold BCA Protein Assay Kit

The Pierce Plant Total Protein Extraction Kit is composed of optimized buffers and filter cartridges that allow for efficient and rapid protein extraction in less than 10 minutes. The kit is designed to rapidly extract denatured or native proteins from plant tissues (leaves, seeds, soft stem, roots, etc.). Simply grind the sample in the lysis buffer in the filter cartridge and centrifuge. The protein extracts can be used for applications such as SDS-PAGE, western blotting, immunoprecipitation, affinity purification, and activity assays.

Related products
Halt Protease & Phosphatase Inhibitor Cocktail
Halt Protease & Phosphatase Inhibitor Cocktail, EDTA-free (100X)
Pierce Rapid Gold BCA Protein Assay Kit
Pierce BCA Protein Assay Kit

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Assay EntryWestern Blot
Quantity50 preps
Shipping ConditionRoom Temperature
Product LinePierce™
Product TypePlant Total Protein Extraction Kit
Unit SizeEach
Contents & Storage
  • Denaturing Lysis Buffer, 25 mL
  • Native Lysis Buffer, 25 mL
  • Protein Extraction Filter Cartridges, 50 Ea.
  • Collection Tubes, 50 Ea.
  • Plastic Rod
croppedsalg3fam

Slide-A-Lyzer  G3 Dialysis Cassettes
Make your dialysis simpler and more secure

Learn more ›

Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link