Informational:Inquire about OEM or Commercial Supply version of this product here.
Invitrogen™

Platinum™ Direct PCR Universal Master Mix

Catalog number: A44647200
Have Questions?
Invitrogen™

Platinum™ Direct PCR Universal Master Mix

Catalog number: A44647200
Have Questions?
Catalog Number
A44647200
Unit Size
4 x 500 reactions
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
100 reactions
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
500 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A446472004 x 500 reactions
-
A44647100100 reactions
-
A44647500500 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

Invitrogen Platinum Direct PCR Universal Master Mix is designed to amplify DNA directly from various samples without the need to purify DNA. It contains high-performing, engineered Platinum II Taq Hot-Start DNA Polymerase with dNTPs in an innovative buffer that enables universal primer annealing for superior performance in direct PCR applications.

Features of Platinum Direct PCR Universal Master Mix include:
• Direct DNA amplification from various samples due to engineered, inhibitor-tolerant Platinum II Taq Hot-Start DNA Polymerase
• Universal primer annealing temperature due to innovative buffer that enables primer annealing at 60°C
• Superior specificity, sensitivity, and yields due to Platinum hot-start technology

Platinum Direct PCR Universal Master Mix is designed to work with a variety of samples of different origins such as animal, human, plant, insect, worm, and bacterial samples. The kit includes optimized reagents to achieve superior results. The quick lysis protocol enables efficient amplification from templates up to 8 kb in size and co-cycling of different length fragments. Samples in the lysis buffer can be stored for later use. Platinum Direct PCR Universal Master Mix is provided with the optional Platinum GC Enhancer for specific amplification and improved yields of GC-rich targets.

Engineered Platinum II Taq DNA polymerase confers faster cycling and tolerance to reaction inhibitors originating from sample material. Proprietary Platinum Taq antibodies block polymerase activity at ambient temperatures and dissociate after the initial denaturation step. This automatic 'hot start' provides increased sensitivity, specificity, and yield, while allowing reaction assembly at room temperature.

Due to the unique composition of the Platinum Direct PCR Universal Master Mix, the annealing temperature of 60°C can be used for most primer pairs that follow the general design rules. Isostabilizing molecules in the buffer increase primer-template duplex stability during the annealing step and contribute to enhanced specificity without the need to optimize annealing temperature for each primer pair. Different PCR assays can be cycled together using the same protocol with a universal primer annealing temperature and an extension step selected for the longest fragment to be amplified.

View more information about Platinum Direct PCR Universal Master Mix ›

Specifications

GC-Rich PCR Performance
High
Polymerase
Platinum II Taq Hot-Start DNA Polymerase
Reaction Speed
Fast
Green Features
Less Hazardous, Less Waste
Product Type
Direct PCR Universal Master Mix
Shipping Condition
Dry Ice
For Use With (Application)
Hot-start PCR, Real Time PCR (qPCR)
Concentration
2X
Fidelity (vs. Taq)
1X
Hot Start
Built-In Hot Start
No. of Reactions
4 x 500 Reactions
Overhang
3'-A
Reaction Format
SuperMix or Master Mix
Size (Final Product)
8 kb

Contents & Storage

Four boxes of Platinum Direct PCR Universal Master Mix, each containing:
• 2X Platinum Direct PCR Universal Master Mix (5 mL)
• Lysis Buffer (2 x 12.5 mL)
• Proteinase K (0.75 mL)
• Platinum GC Enhancer (2 x 1.25 mL)
• Nuclease-free water (5 mL)

Store at –20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text