MagMAX™ Cell and Tissue DNA Extraction Buffer
MagMAX™ Cell and Tissue DNA Extraction Buffer
Applied Biosystems™

MagMAX™ Cell and Tissue DNA Extraction Buffer

MagMAX DNA Cell and Tissue Extraction Buffer, a component of the MagMAX DNA Multi-Sample Ultra 2.0 Kit with Cell andRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
A45469100 Preps
A454701000 Preps
Catalog number A45469
Price (USD)
129.65
Online Exclusive
138.00
Save 8.35 (6%)
Each
-
Add to cart
No. of Reactions:
100 Preps
Recurring order eligible. Learn more »
Delivery information
Standard Products - 1-2 business days for Beijing, Shanghai, Guangzhou, Shenzhen and provincial capitals, 2-3 business days for 2nd-tier cities, and 4 business days for 3rd-tier cities and remote areas.
Air Freight Restricted Products - 2-4 business days for Beijing, Shanghai, Guangzhou, Shenzhen and provincial capitals, 3-5 business days for 2nd-tier cities, and 6-10 business days for 3rd-tier cities and remote areas.
Supply Center: Delivered from the Supply Center closest to you.
Price (USD)
129.65
Online Exclusive
138.00
Save 8.35 (6%)
Each
Add to cart
Ask our AI about this Product

MagMAX DNA Cell and Tissue Extraction Buffer, a component of the MagMAX DNA Multi-Sample Ultra 2.0 Kit with Cell and Tissue Extraction Buffer, is offered separately here for those kit users who need additional buffer.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Next-Generation Sequencing, PCR, Real-Time PCR
High-throughput CompatibilityAutomated Protocols, High-throughput Compatible, Manual Protocols
No. of Reactions100 Preps
Product LineMagMAX™
Product TypeCell and Tissue DNA Extraction Buffer
Quantity100 Preps
Sample TypeCells, Tissue
Shipping ConditionRoom Temperature
TargetGenomic DNA
Unit SizeEach
Contents & Storage
60 mL Cell and Tissue DNA Extraction Buffer
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

浏览此商品的顾客同时也查看了



Documents & Downloads

Certificates

Frequently asked questions (FAQs)

Yes, we have protocols for using the MagMAX DNA Multi-Sample Ultra 2.0 Kit (Cat. No. A36570) to isolate genomic DNA from these sample types, but they require purchase of an additional Cell and Tissue Extraction Buffer. The kit and buffer can be purchased together as Cat. No. A45721 or the Cell and Tissue Extraction Buffer can be purchased as a stand alone item (Cat. No. A45469 or A45470).

The suggested workflow for bone marrow aspirates is as follows:
- Use only up to 200 µL of bone marrow aspirate
- Centrifuge at 200 x g for 10 mins
- Remove the supernatant (leaving about 50 µL of aspirate with the loose pellet) and resuspend the cell pellet in 400 µL of Cell and Tissue Extraction Buffer
- From this point, follow the protocol for isolation of DNA from buffy coat samples

The suggested workflow for 1-2.0 x 10E6 PBMCs is as follows:
- Centrifuge at 500-800 x g for 10 mins
- Remove the supernatant (leaving the cell pellet) an resuspend the cell pellet in 400 L of Cell and Tissue Extraction Buffer
- From this point, follow the protocol for isolation of DNA from cultured cell samples

Yes, we have protocols for using the MagMAX DNA Multi-Sample Ultra 2.0 Kit (Cat. No. A36570) to isolate DNA from cells and tissue, but they require purchase of an additional Cell and Tissue Extraction Buffer. The kit and buffer can be purchased together as Cat. No. A45721 or the Cell and Tissue Extraction Buffer can be purchased as a stand alone item (Cat. No. A45469 or A45470).

RNase A digestion is optional, but we highly recommend doing it. We recommend using PureLink RNase A (20 mg/mL) (Cat. No. 12091021).

其他推荐产品



Share catalog number, name or link

1x1 image pixel for data collection