Ion Torrent™

Oncomine™ BCR IGH SR Assay, DNA

Catalog number: A45483
Have Questions?
Ion Torrent™

Oncomine™ BCR IGH SR Assay, DNA

Catalog number: A45483
Have Questions?
Catalog Number
A45483
Unit Size
24 reactions
Price (USD)
Quantity
Catalog NumberUnit SizePrice (USD)Quantity
A4548324 reactions
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Ion Torrent Oncomine BCR IGH SR Assay, DNA, is a robust, targeted next-generation sequencing (NGS) assay designed for use in hematology-oncology and minimal residual disease (MRD) monitoring research. It can also be used to investigate immunotherapy biomarkers, as well as in infectious and autoimmune disease research. The assay kit includes a single pool of multiplex PCR primers, total dNTPs, and library reagents.

The Oncomine BCR IGH SR Assay, DNA, accurately identifies and measures clonal expansion of B lymphocytes using genomic DNA from bone marrow, whole blood, peripheral blood leukocytes (PBLs), peripheral blood mononuclear cells (PBMCs), and fresh-frozen and formalin-fixed paraffin-embedded (FFPE) research samples. Using Ion AmpliSeq technology, the assay identifies unique B-cell clones through targeting of the highly diverse complementarity-determining region 3 (CDR3) of the B-cell receptor (BCR) immunoglobulin heavy (IGH) chain from a DNA template. The nucleotide sequence of the IGH CDR3 region serves as a natural barcode to enable clone identification for MRD clinical research. The measurements of B-cell clonal expansion and diversity are useful for pharmacodynamics and vaccine biomarker research. IGH CDR3-region amino acid motifs may reveal signatures of B-cell responses to defined antigens for use as future markers of autoimmune or infectious disease. For RNA samples, please see Cat. No. A45484.

Benefits of the Oncomine BCR IGH SR Assay, DNA, include:
• High sensitivity and exceptionally low limit of detection for rare clone identification via dual-barcode indexing and high template capture efficiency of Ion AmpliSeq multiplex PCR
• Compatibility with extracted DNA from an array of sample types, including fresh-frozen and FFPE tissue, bone marrow, whole blood, PBLs, and PBMCs
• Assay primers designed to amplify all variable and joining gene alleles in gold-standard IMGT database
• Generation of large libraries with minimal input requirement, facilitating study of rare clones
• Detection of clones of interest at a frequency as low as 10-6
• Efficient workflow with 48-hour library-to-results time
• Flexible input requirements ranging from 50 ng to 2 μg
• Streamlined and user-friendly informatics solution with automated clonotyping, clonal lineage analysis, reporting of key repertoire features, and multi-sample analysis capability to generate MRD analysis for research samples
• Ultra-low substitution error rate minimizing artifacts arising from sequencing errors, enabling highly accurate assessment of clonal heterogeneity in malignant B cells of interest in research samples

The Oncomine BCR IGH SR Assay, DNA, uses multiplex PCR primers to generate 90-bp amplicons that can be sequenced on all chip types supported by the Ion GeneStudio S5 sequencing systems, allowing you to pick the best multiplexing configuration for your unique sample batching needs and throughput requirements. The entire workflow from library preparation to analysis of samples can be accomplished in two days using the Ion Chef templating system and Ion GeneStudio S5 sequencing system.

Learn more about the assay ›

Note: Information about data analysis using Ion Reporter Software v5.12 can be found in the User Guide below.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Sample Type
DNA
Sequencing Type
Genome and DNA Sequencing
For Use With (Application)
Sequencing
Libraries
Targeted Sequencing Library
No. of Reactions
24
Product Line
Oncomine™
Product Type
BCR IGH SR Assay
Shipping Condition
Approved for shipment on Wet or Dry Ice
Workflow Step
Library Generation

Contents & Storage

• Oncomine BCR IGH-SR Assay, DNA, store in freezer (-5 to -30°C)
• Ion AmpliSeq Library Kit Plus, store in freezer (-5 to -30°C)
• dNTP Mix, 25 mM each, store in freezer (-5 to -30°C)

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text