Ion Torrent™

Ion AmpliSeq™ Mouse TCR Beta SR Assay, DNA

Catalog number: A45488
Have Questions?
Ion Torrent™

Ion AmpliSeq™ Mouse TCR Beta SR Assay, DNA

Catalog number: A45488
Have Questions?
Catalog Number
A45488
Unit Size
24 reactions
Price (USD)
Quantity
Catalog NumberUnit SizePrice (USD)Quantity
A4548824 reactions
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Ion AmpliSeq Mouse TCR Beta SR Assay is a robust, targeted next-generation sequencing (NGS) assay designed to accurately identify and measure the clonal expansion of T lymphocytes by targeting the complementarity-determining region 3 (CDR3) of the T-cell receptor (TCR) gene locus from gDNA input. The assay can be used for basic and translational research to identify T-cell clones since the nucleotide sequence of the CDR3 region is unique to each T cell and codes for the part of the TCR beta chain that is involved in antigen recognition. For RNA samples, please see Cat. No. A45489.

The high sensitivity and specificity of the assay enables key applications in immunology, immuno-oncology, and vaccine research. The high-sensitivity dual-barcode indexing, flexible input requirements (100 ng–1 μg), high-depth sequencing, and high-throughput capability make this assay ideal for testing a variety of use cases in mouse models, including the role of T cells in generating immune response to immunotherapies such as checkpoint blockade inhibitors, cancer vaccines, or chimeric antigen receptor (CAR) T-cell therapies.

Benefits of the Ion AmpliSeq Mouse TCR Beta SR Assay, DNA, include:
Compatibility with a vast array of research sample types, including fresh-frozen and FFPE tissue, whole blood, peripheral blood leukocytes, and peripheral blood mononuclear cells
High sensitivity and low limit of detection for rare clone identification through dual-barcode indexing
Efficient workflow with 48-hour sample-to-results time
Flexible input requirements ranging from 100 ng to 1 μg
Streamlined and user-friendly informatics solution with multi-sample analysis functionality

The Ion AmpliSeq TCR Beta-SR Mouse Kit, DNA, provides a single pool of multiplex PCR primers and library reagents to generate 90-bp amplicons that can be sequenced on all chip types supported by the Ion GeneStudio S5 sequencing systems, allowing you to pick the best multiplexing configuration for your unique sample batching needs and throughput requirements. The entire workflow from library preparation to analysis of samples can be accomplished in two days using the Ion Chef templating system and the Ion GeneStudio S5 sequencing system.

Learn more about the assay ›

Note: Information about data analysis using Ion Reporter Software v5.12 can be found in the User Guide below.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Sample Type
DNA
Sequencing Type
Genome and DNA Sequencing
For Use With (Application)
Sequencing
Libraries
Targeted Sequencing Library
No. of Reactions
24
Product Line
AmpliSeq™
Product Type
Mouse TCR Beta SR Assay
Shipping Condition
Approved for shipment on Wet or Dry Ice
Species
Mouse
Workflow Step
Library Generation

Contents & Storage

• Ion AmpliSeq Mouse TCRB-SR Assay, DNA, store in freezer (-5 to -30°C)
• Ion AmpliSeq Library Kit Plus, store in freezer (-5 to -30°C)
• dNTP Mix, 25 mM each, store in freezer (-5 to -30°C)

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text