Ion Torrent™

Ion AmpliSeq™ Mouse TCR Beta SR Assay, RNA

Catalog number: A45489
Have Questions?
Ion Torrent™

Ion AmpliSeq™ Mouse TCR Beta SR Assay, RNA

Catalog number: A45489
Have Questions?
Catalog Number
A45489
Unit Size
24 reactions
Price (USD)
Quantity
Catalog NumberUnit SizePrice (USD)Quantity
A4548924 reactions
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Ion AmpliSeq Mouse TCR Beta SR Assay is a robust, targeted next-generation sequencing (NGS) assay designed to accurately identify and measure the clonal expansion of T lymphocytes by targeting the complementarity-determining region 3 (CDR3) of the T-cell receptor (TCR) gene locus from total RNA input. The assay can be used for basic and translational research to identify T-cell clones since the nucleotide sequence of the CDR3 region is unique to each T cell and codes for the part of the TCR beta chain that is involved in antigen recognition. For DNA samples please see Cat. No. A45488.

Benefits of the Ion AmpliSeq Mouse TCR Beta SR Assay, RNA, include:
Compatibility with a vast array of research sample types, including FFPE tissue, fresh-frozen (FF) tissue, whole blood, PBLs, and PBMCs
High sensitivity and low limit of detection (LoD) for rare clone identification through dual-barcode indexing
Efficient workflow with 48 hour sample-to-results time
Flexible input requirements ranging from 25 ng to 1 μg
Streamlined and user-friendly informatics solution with with multi-sample analysis functionality

The Ion AmpliSeq Mouse TCR Beta SR Kit, RNA, provides a single pool of multiplex PCR primers and library reagents to generate 90-bp amplicons that can be sequenced on all chip types supported by the Ion GeneStudio S5 sequencing systems, allowing you to pick the best multiplexing configuration for your unique sample batching needs and throughput requirements. The entire workflow from library preparation to analysis of samples can be accomplished in two days using the Ion Chef templating system and Ion GeneStudio S5 system.

The Ion AmpliSeq Mouse TCR Beta SR RNA Assay, with its high sensitivity and specificity, supports key applications in the field of immunology, immuno-oncology, hemato-oncology, and vaccine research. The high-sensitivity dual-barcode indexing, flexible input requirements (100 ng–1 μg), high-depth sequencing, and high-throughput capability make this assay ideal for testing a variety of hypotheses for basic or translational biomarker research. A small fragment size requirement from the input materials for library creation and high multiplexing on the Ion 530 Chip of the Ion GeneStudio S5 sequencers enables researchers to focus on testing a variety of use cases in mouse models including the role of T cells in generating immune response to immunotherapies such as checkpoint blockade inhibitors, cancer vaccines, or chimeric antigen receptor (CAR) T-cell therapies.

Learn more about the assay ›

Note: Information about data analysis using Ion Reporter Software v5.12 can be found in the User Guide below.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Sample Type
RNA (Total)
Sequencing Type
Genome and DNA Sequencing
For Use With (Application)
Sequencing
Libraries
Targeted Sequencing Library
No. of Reactions
24
Product Line
AmpliSeq™
Product Type
Mouse TCR Beta SR Assay
Shipping Condition
Approved for shipment on Wet or Dry Ice
Species
Mouse
Workflow Step
Library Generation

Contents & Storage

• Ion AmpliSeq TCRB-SR Assay, RNA (-5 to -30°C)
• Ion AmpliSeq Library Kit Plus (-5 to -30°C)
• Ion NGS RT Kit (-5 to -30°C)

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text