Thermo Scientific™

SuperSignal™ Western Blot Substrate Trial Pack, Atto & Pico PLUS

Catalog number: A45918
Have Questions?
Thermo Scientific™

SuperSignal™ Western Blot Substrate Trial Pack, Atto & Pico PLUS

Catalog number: A45918
Have Questions?
Catalog Number
A45918
Unit Size
20 mL + 20 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A4591820 mL + 20 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
This substrate trial pack contains sample sizes of two substrates: SuperSignal West Atto Ultimate Sensitivity Chemiluminescent Substrate and SuperSignal West Pico PLUS Chemiluminescent Substrate to enable trial use of the most recommended substrates for routine and high-sensitivity western blotting.

SuperSignal West Atto Ultimate Sensitivity Chemiluminescent Substrate
Thermo Scientific SuperSignal West Atto Ultimate Sensitivity Chemiluminescent Substrate is an ultra-sensitive enhanced chemiluminescent (ECL) substrate for high attogram protein level detection by western blot analysis with horseradish peroxidase (HRP) enzyme. It is designed for detection of very low-abundant targets or the maximum levels of sensitivity required when using precious samples or costly antibodies.

• Sensitivity: low-femtogram to high-attogram
• Stability: eight hours for working solution, one year for kit at 4°C
• Compatibility: nitrocellulose and PVDF membranes
• Signal duration: up to four hours
• Recommended primary antibody concentration: 1:1,000–1:5,000 dilution (0.2–1.0 μg/mL)
• Recommended secondary antibody concentration: 1:100,000–1:250,000 dilution (4–10 ng/mL)

View full product information ›

SuperSignal West Pico PLUS Chemiluminescent Substrate
Thermo Scientific SuperSignal West Pico PLUS Chemiluminescent Substrate enables picogram to high femtogram-level protein detection by western blot analysis. It is compatible with different membranes, blocking reagents, and a wide range of antibody dilutions, making it an ideal choice for routine western blotting applications.

• Sensitivity: picogram to femtogram
• Stability: eight hours for working solution; one year for kit at room temperature
• Compatibility: nitrocellulose and PVDF membranes
• Signal duration: 6 to 24 hours
• Recommended primary antibody concentration: 1:1,000–1:5,000 dilution (0.2–1.0 μg/mL)
• Recommended secondary antibody concentration: 1:20,000–100,000 dilution (10–50 ng/mL)

View full product information ›

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
Substrate
Detection Method
Chemiluminescent
Form
Liquid
For Use With (Application)
Western Blot
Membrane Compatibility
Nitrocellulose, PVDF
Product Line
SuperSignal™
Quantity
Trial Pack
Substrate
Horseradish Peroxidase
Sensitivity
Picogram to femtogram, low-femtogram to high-attogram
Sufficient For
4 Mini-Gel Blots, 400 sq. cm of Membrane
Shipping Condition
Approved for shipment at Room Temperature or on Wet or Dry Ice

Contents & Storage

• 20 mL SuperSignal West Pico PLUS reagent, store at room temperature
• 20 mL SuperSignal West Atto reagent, store at 2–8°C

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text