QuantStudio™ Pro Block Upgrade Kit, 96-well, 0.1 mL
Copy product name
Copy catalog number
Copy catalog number and product name
Copy link
Applied Biosystems™
QuantStudio™ Pro Block Upgrade Kit, 96-well, 0.1 mL
The QuantStudio Pro Block Upgrade Kit is an optional accessory for the QuantStudio Pro Real-Time qPCR System. Interchangeable blocks enableRead more
Have Questions?
Catalog Number
Quantity
A45955
1 kit
Catalog number A45955
Price (USD)
-
Request A Quote
Quantity:
1 kit
The QuantStudio Pro Block Upgrade Kit is an optional accessory for the QuantStudio Pro Real-Time qPCR System. Interchangeable blocks enable application versatility with easy, powered-up interchange of block types.
The QuantStudio 7 Pro system accommodates the interchange of 96-well (0.2 mL), 96-well (0.1 mL), 384-well, and TaqMan array card (TAC) blocks. The system is delivered with one block type; other block types may be purchased separately. For a more value-priced system that does not support TaqMan array cards or automation (but that can be upgraded to do so at a later date), please see our QuantStudio 6 Pro Real-Time PCR System. And for the ultimate in flexibility, consider the QuantStudio 12K Flex Real-Time PCR System, which also accommodates TaqMan OpenArray plates.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Real Time PCR (qPCR)
For Use With (Equipment)QuantStudio™
Product LineQuantStudio™
Quantity1 kit
Product TypeOther Accessories
Unit SizeEach
Contents & Storage
96-well, 0.1 mL block, heated cover and accessories