Search Thermo Fisher Scientific
Catalog Number | Unit Size | Price (USD) | |
---|---|---|---|
A46731 | 2 runs | 2,070.00 |
The Ion Torrent GX7 Chip and Genexus Coupler includes two GX7 chips and two Genexus couplers, sufficient for two initializations of the Genexus Integrated Sequencer. The GX7 Chip generates 20–25 million sequencing reads per lane, or 80–100 million reads per chip. The four-lane chip design supports maximum flexibility for sample processing, enabling sequencing of one to four lanes per run, with the in-use stability to utilize any remaining lanes within two weeks of the initial run.
As part of the Genexus Integrated Sequencer workflow, the GX7 Chip:
• Achieves 20–25 million reads per lane for 200 base-read libraries
• Maximizes flexibility with the ability to utilize one to four chip lanes per run
• Enables parallel processing of up to four compatible assays in a single run
• Supports intake variability of clinical research samples
The Genexus Integrated Sequencer automates library preparation, sequencing, analysis, and reporting in a hands-off, walkaway workflow to deliver results in a single day. The innovative, multilane sequencing chip is designed to enable cost-effective processing of 1 to 32 single pool libraries.