The Applied Biosystems QuantStudio 5 Dx Real-Time PCR System is an in vitro diagnostic (IVD) quantitative PCR system that delivers proven performance and support to help meet the needs of clinical/hospital laboratories and test developers, in a compact footprint and cost-effective package. This system is both US-IVD and CE-IVD, IVDR compliant.
Flexibility and securityThe QuantStudio 5 Dx software is a combined-function software that enables both Development (DEV) and In Vitro Diagnostic (IVD) operation as discrete operating modes, minimizing risk of operation in the incorrect instance.
The software enables you to maintain secure access to the instrument, specify user access to various software functions, record all actions completed by users, and sign data electronically by virtue of Security, Auditing, and E-signature features.
This QuantStudio 5 Dx system is offered in a 96-well, 0.2 mL configuration and accommodates reaction volumes from 10 to 100 uL, as well as both standard and fast chemistries to meet varying user demands.
Trusted performanceResulting from over 20 years of real-time PCR experience, the QuantStudio 5 Dx system includes proven Applied Biosystems OptiFlex technology and VeriFlex blocks for enhanced data quality. By enabling de-coupling of excitation and emission filters with OptiFlex technology, users may optimize performance for specific dyes and when multiplexing. With VeriFlex blocks, users may precisely set the temperature on six independently-controlled thermal zones across the 96-well sample block for optimal PCR amplification performance.
Cost effectiveThe QuantStudio 5 Dx system is our most affordable IVD qPCR platform. It is factory-calibrated and, following installation qualification and operational qualification by a factory-certified engineer, ready for operation by trained users. Subsequent IVD dye calibrations are now user-enabled for convenience and to minimize ongoing operating costs.
More information about the QuantStudio 5 Dx Real-Time PCR System ›Learn more about available financing options and programs ›For In Vitro Diagnostic Use.