The CLA (Cell Line Authentication) IdentiFiler Plus PCR Amplification Kit simplifies the amplification step during authentication of cell lines. It includes the same primers and allelic ladder as the widely used AmpFLSTR Identifiler Plus kit and harnesses next-generation PCR amplification technology to help provide enhanced sensitivity, a cleaner baseline, improved performance with mixtures, and the ability to overcome high levels of PCR inhibition.
Features of the CLA IdentiFiler Plus PCR Amplification Kit include: • 5-dye, 16-locus STR kit compatible with all cell line databasing loci standards • Improved version of legacy AmpFLSTR Identifiler Plus kit • Compatible with SeqStudio, 3730/xL, and 3500/xL genetic analyzers
Cell line authentication using short tandem repeat (STR) analysis kits can be used both for human sample authentication (HSA) and mixed sample analysis (MSA). In HSA, there is a need to verify the DNA profile of the sample to check for sample cross-contamination or sample mix-up, or to perform a quality check. For MSA there is a need to deconvolute the presence of multiple contributors in a sample (chimerism) through identification of multiple DNA genotypes in the sample.
SNP Genotyping (Fragment Analysis-Based), STR (Short Tandem Repeat) Analysis
Genotyping Target
SNPs (Unknown or Numerous)
Product Type
Plus PCR Amplification Kit
Shipping Condition
Wet Ice
Species
Human
Concentration
AmpFlSTR Identifiler Plus Master Mix - 1 tube at 0.5 mL, AmpFlSTR Control DNA 9947A - 1 tube at 0.3 mL, AmpFlSTR Identifiler Plus Primer Set - 1 tube at 0.25 mL, AmpFlSTR Identifiler Plus Allelic Ladder - 1 tube at 0.025 mL
No. of Reactions
50 Reactions
Volume
0.5 mL
Unit Size
50 reactions
Contents & Storage
• 1 AmpFLSTR Identifiler Plus PCR Amplification Kit, store at -5 to -30°C