The Gibco AAV-MAX Control Plasmids Kit consists of an optimized mix of three plasmids needed to produce adeno-associated virus serotypeRead more
Have Questions?
Catalog Number
Quantity
A47672
1 Kit
Catalog number A47672
Price (USD)
636.00
Each
-
Add to cart
Quantity:
1 Kit
Price (USD)
636.00
Each
Add to cart
Ask our AI about this Product
The Gibco AAV-MAX Control Plasmids Kit consists of an optimized mix of three plasmids needed to produce adeno-associated virus serotype 2 (AAV2)−expressing GFP: pAAV-CMV-emGFP, pAAV-Rep2Cap2, and pHelper. These three plasmids must be used with a HEK293 cell line that stably expresses the adenovirus E1 gene. The kit contains an optimized mix of the three plasmids, as well as a separate tube of each plasmid, providing users with flexibility to change plasmid ratios if desired.
• Saves process development time with optimized plasmid ratios • Serves as a positive control for AAV2-GFP production with the AAV-MAX Helper-Free AAV Production System (Cat. No. A51217)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration1 mg/mL
Delivery TypeTransfection
Expression SystemMammalian
FormatKit
Product LineAAV-MAX
Product TypePlasmid
Quantity1 Kit
Regulatory StatusResearch Use Only (RUO)
Shelf Life2 Years
Shipping ConditionIce
Unit SizeEach
Contents & Storage
• 1 tube containing mixture of all three plasmids, 1.5 mL • 1 tube of each of the three plasmids alone, 0.75 mL
Store at –20°C.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our
Privacy Notice.
Kunden, die diesen Artikel ansahen, interessierten sich auch für