Applied Biosystems™

TaqMan™ SARS-CoV-2, Flu A/B, RSV RT-PCR Assay Kit

Catalog number: A47702
Have Questions?
Applied Biosystems™

TaqMan™ SARS-CoV-2, Flu A/B, RSV RT-PCR Assay Kit

Catalog number: A47702
Have Questions?
Catalog Number
A47702
Unit Size
1000 reactions
Price (USD)
Catalog NumberUnit SizePrice (USD)
A477021000 reactions
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Detect RNA from multiple respiratory viruses simultaneously with the TaqMan SARS-CoV-2, FluA/B, RSV RT-PCR Assay Kit, a highly accurate, real-time PCR multiplex assay that allows you to expand your respiratory sample analysis menu while maintaining low operational costs.

Features of the TaqMan SARS-CoV-2, FluA/B, RSV RT-PCR Assay Kit include:
Single assay for SARS-CoV-2, influenza A/B, and RSV RNA—detect and differentiate between infection states and get the information needed to help analyze distribution of SARS-CoV-2, flu, and RSV nucleic acid
Affordable and scalable—add to your existing SARS-COV-2 assay menu and infrastructure to expand your respiratory sample analysis while maintaining low operational costs and workflow simplicity
Sensitive and specific—get 100% homology to >99.9% of known SARS‐CoV‐2 genomes* and robust performance for detecting flu A/B and RSV RNA, providing increased confidence in results
Complete solution for labs—Applied Biosystems Design & Analysis Software automatically converts genetic analysis data into a readable report, helping reduce risk of user interpretation error

Other reagents recommended to be used in conjunction with the TaqMan SARS-CoV-2, FluA/B, RSV RT-PCR Assay Kit include:
TaqMan SARS-CoV-2, Flu A/B, RSV RNA Control
TaqMan SARS-CoV-2 Control Dilution Buffer
TaqPath 1-Step Multiplex Master Mix (No ROX)
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
Primer-probe
Product Line
TaqMan
Quantity
1000 Reactions
Shipping Condition
Dry Ice
Software
Applied Biosystems™ QuantStudio™ Design and Analysis Software 2.4.x
Target Organism Class
SARS-CoV-2 (S & N genes), Influenza A/B, RSV

Contents & Storage

Store at -30°C to -10°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text