Applied Biosystems™

TaqMan™ SCID/SMA Assay

Catalog number: A47929
Have Questions?
Applied Biosystems™

TaqMan™ SCID/SMA Assay

Catalog number: A47929
Have Questions?
Catalog Number
A47929
Unit Size
20,000 reactions
Price (USD)
Catalog Number
Unit Size
4,000 reactions
Price (USD)
Catalog Number
Unit Size
8,000 reactions
Price (USD)
Catalog NumberUnit SizePrice (USD)
A4792920,000 reactionsRequest A Quote
A479274,000 reactionsRequest A Quote
A479288,000 reactionsRequest A Quote
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Applied Biosystems TaqMan SCID/SMA Assay is a multiplex real-time PCR assay that detects the SMN1 gene and TREC, as well as the RNase P gene as an internal genomic control. For an assay that also detects KREC, please see the TaqMAN SCID/SMA Plus Assay (Cat. No. A48566).

Features of the TaqMan SCID/SMA Assay include:
• Optimized for SCID and SMA
• Direct extraction from DBS samples for reduced hands-on time and cost
• Compatible with 96- or 384-reaction protocols
• High-quality results for 96 samples within 55 min using the fast cycling protocol
• Minimal liquid-dispensing steps for easy automation

Complete solution from DBS to result
The TaqMan SCID/SMA Assay detects SMN1, TREC, as well as the RNase P gene as an internal genomic control. The assay solution contains a mixture of primers and probes for each target plus an SMN2 blocker. From DBS to result, less than 40 min of hands-on time and only four reagent-dispensing steps are required for 96-well plates. One additional pipetting transfer step from DBS lysate extraction plate to qPCR plate is needed. A real-time PCR instrument with at least five filters is required to detect the reporter dyes plus the passive reference dye.

The TaqMan SCID/SMA Assay is a multiplex real-time PCR assay that detects exon 7 of SMN1 and TREC. The assay includes the RNase P gene as an internal amplification control, verifying successful sample extraction. The assay is optimized for high specificity to SMN1, eliminating cross-detection of SMN2.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
Primer-probe
For Use With (Equipment)
QuantStudio™ 12 k Flex, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7, QuantStudio™ Dx System (in RUO Mode)
Product Line
TaqMan™
Shipping Condition
Dry Ice
No. of Reactions
20,000 Reactions
Sample Type
DNA (Genomic), cDNA

Contents & Storage

• DNA Extract All Reagents Kit
• TaqPath ProAmp Multiplex Master Mix
• TaqMan SCID/SMA Assay

Upon receipt, store at -20°C. After opening, store at 4°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text