TrueMark™ Respiratory Panel 2.0 Amplification Control
TrueMark™ Respiratory Panel 2.0 Amplification Control
Applied Biosystems™

TrueMark™ Respiratory Panel 2.0 Amplification Control

The TrueMark Respiratory Panel 2.0 Amplification Control contains a linearized multi-target plasmid with target sequences for each respiratory tract microbiotaRead more
Have Questions?
Catalog NumberQuantity
A481015 x 50 μL
Catalog number A48101
Price (EUR)
794,00
Each
-
Add to cart
Quantity:
5 x 50 μL
Recurring order eligible. Learn more »
Price (EUR)
794,00
Each
Add to cart
Ask our AI about this Product
The TrueMark Respiratory Panel 2.0 Amplification Control contains a linearized multi-target plasmid with target sequences for each respiratory tract microbiota assay on the TrueMark Respiratory Panel 2.0, OpenArray plate and TrueMark Respiratory Panel 2.0, TaqMan array card. The control can be included in respiratory tract microbiota profiling experiments as a positive control for panel-specific amplification.

The plasmid also contains target sequences for TaqMan Universal RNA Spike In/Reverse Transcription (Xeno) Control, TaqMan Universal Extraction Control Organism (B. atrophaeus), and the human RNase P RPPH1 gene. In addition, the plasmid contains target sequences for additional qualified TaqMan assays for respiratory tract microbiota profiling that may be included on custom arrays or cards. Refer to the Respiratory Tract Microbiota Profiling Experiments v2 using TaqMan Array Cards Application Guide or Respiratory Tract Microbiota Profiling Experiments v2 using OpenArray Application Guide for the full assay list.

The TrueMark Respiratory Panel 2.0 Amplification Control is supplied at a concentration of 1 x 105 copies/μL. During real-time PCR, the control can be used as a stand-alone sample to verify assay performance and to help with troubleshooting.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration1 x 105 copies/μL
For Use WithTrueMark™ Respiratory Panel 2.0, TaqMan™ Array Card, TrueMark™ Respiratory Panel 2.0, OpenArray™ Plate
FormatTube
Product LineTrueMark™
Quantity5 x 50 μL
Shipping ConditionDry Ice
Volume250 μL
Detection MethodPrimer-probe
Unit SizeEach
Contents & Storage
Store at -25°C to -15°C.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3180587Certificate of AnalysisFeb 28, 2025A48101
2811263Certificate of AnalysisJan 29, 2025A48101
2811270Certificate of AnalysisJan 29, 2025A48101
2802707Certificate of AnalysisJan 29, 2025A48101
2822291Certificate of AnalysisJan 29, 2025A48101
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection