Informational:Discover the future of Gibco Media selection with our upcoming updated experience. View details >
Gibco™

Viral Production Medium

Catalog number: A4817901
Have Questions?
Gibco™

Viral Production Medium

Catalog number: A4817901
Have Questions?
Catalog Number
A4817901
Unit Size
1 L
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
6 x 1 L
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
10 L
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A48179011 L
-
A48179026 x 1 L
-
A481790310 L
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Gibco Viral Production Medium is a chemically defined, serum-free, protein-free medium, that is animal origin-free and specially developed for growth and transfection of Viral Production Cells 2.0 (suspension-adapted HEK 293F-derived cells) for adeno-associated virus (AAV) production as part of the AAV-MAX Helper-Free AAV Production System.

Features of Viral Production Medium include:
• Supports growth of suspension Viral Production Cells 2.0 to densities of over 1.2 x 107 cells/mL
• Enables production of high AAV titers, greater than 5 x 1010 vg/mL (unpurified)
• Supports growth and transfection of Viral Production Cells 2.0 from shake flask to bioreactor scales

Viral Production Medium requires supplementation with 4 mM GlutaMAX Supplement (Cat. No. 35050038) prior to use.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Line
Viral Production Cells 2.0
Cell Type
HEK 293 cells
Classification
Serum-free, Animal Origin-free, Chemically-defined, Protein-free
Culture Type
Suspension Cell Culture
Expression System
Mammalian
Form
Liquid
Product Type
AAV Production Medium
Shelf Life
12 months from date of manufacture
Sterility
Sterile
With Additives
Glucose, Sodium Pyruvate, Sodium Pyruvate, GlutaMAX
Without Additives
No Glutamine
Format
Bottle
For Use With (Application)
AAV Production
pH
7
Product Line
AAV-MAX
Quantity
1 L
Regulatory Status
RUO - Research Use Only
Shipping Condition
Room Temperature

Contents & Storage

1 L bottle; store at 2–8°C, protected from light

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text