Applied Biosystems™

MagMAX™ Viral/Pathogen II (MVP II) Nucleic Acid Isolation Kit

Catalog number: A48383
Have Questions?
Applied Biosystems™

MagMAX™ Viral/Pathogen II (MVP II) Nucleic Acid Isolation Kit

Catalog number: A48383
Have Questions?
Catalog Number
A48383
Unit Size
Up to 2,000 preps
Price (USD)
Quantity
Catalog NumberUnit SizePrice (USD)Quantity
A48383Up to 2,000 preps
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The MagMAX Viral/Pathogen II Nucleic Acid Isolation Kit (CE-IVD marked) is for use by qualified and trained clinical laboratory personnel specifically instructed and trained in the techniques of magnetic bead purification, either manual or automated, and in vitro diagnostic procedures. The kit is specifically designed to recover viral RNA and DNA from human nasopharyngeal swabs and RNA from saliva specimens utilizing MagMAX magnetic-bead technology for reproducible recovery of high-quality nucleic acid.

This product is approved under the In Vitro Diagnostic Regulations (IVDR) in the European Union.

Features of the MVP II kit include:
• Compatible with multiple downstream assays
• Automation-ready protocols designed for KingFisher Flex magnetic particle processor
• Fast procedure allows for 96 samples to be processed in less than 30 minutes
• Flexible protocol accommodates sample volume inputs of 200 or 400 μL of transfer medium
• Elution volume of 50 μL

Access the Instructions for Use approved for your country ›

Intended use statement
The MagMAX Viral/Pathogen II Nucleic Acid Isolation Kit is a nucleic acid purification kit based on magnetic bead technology. The kit is intended for the isolation and purification of viral nucleic acids from human nasopharyngeal swabs and viral RNA from saliva specimens, and is suitable for use in molecular detection by qPCR.

The kit is intended for use by qualified and trained clinical laboratory personnel specifically instructed and trained in the techniques of magnetic bead purification, either manual or automated, and in vitro diagnostic procedures.

Disclaimer
*The 24 months shelf life applies in select countries only and availability is subject to terms and conditions. Please check with your country specific representative for more information
For In-Vitro Diagnostic Use.

Specifications

Format
96 Deep-well Plate
Purification Time
30 min.
Sample Type
Nasopharyngeal Swabs and Saliva
High-throughput Compatibility
High Throughput-Compatible
For Use With (Application)
Real-Time PCR
No. of Reactions
2000 Preps
Product Line
MagMAX™
Shelf Life
24 months*
Shipping Condition
Room Temperature
Target
Viral DNA/RNA

Contents & Storage

Now available in a convenient single-box format with a single expiration date.

• 550 mL Binding Solution
• 1,000 mL Wash Solution
• 20 mL MVP II Binding Beads
• 10 mL Proteinase K
• 100 mL Elution Buffer

Store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text