CTS™ Rotea™ Hi-Flow Single-Use Kit
CTS™ Rotea™ Hi-Flow Single-Use Kit
Thermo Scientific™

CTS™ Rotea™ Hi-Flow Single-Use Kit

The Gibco CTS Rotea Hi-Flow Single-Use Kit is part of the Gibco CTS Rotea Counterflow Centrifugation System. The kit isRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A492395 Kits
A4657510 Kits
2 Options
Catalog number A49239
Price (USD)/ 5 kits
-
Request A Quote
Quantity:
5 Kits
The Gibco CTS Rotea Hi-Flow Single-Use Kit is part of the Gibco CTS Rotea Counterflow Centrifugation System. The kit is capable of flow rates of 30-160 mL/min. Each kit can perform a wide range of cell processing steps, making ideal for wash & concentrate, separation, isolation, and more. The single-use kit provides flexibility for process development and easily supports commercial scale manufacturing. Each kit is compatible with sterile luer connectors or thermal welding.

Tubing material: DEHP-free PVC
Tube dimensions: 0.160 in. OD, 0.116 in. ID
Pump tube material: Bioprene™ plastic

Kits are gamma-irradiated and individually packaged.

Specifications
MaterialTritan MX711 Copolyester (Chamber), DEHP-free PVC (Tubing)
For Use With (Equipment)CTS Rotea Counterflow Centrifugation System
DisposableSingle-use
Capacity20 L
Max. RCF3000 x g
Certifications/ComplianceISO 13485
Quantity5 Kits
Sterilization MethodGamma-irradiated
Cell TypeAgnostic
For Use With (Application)Cell and Gene Therapy Research, Development, and Manufacturing
Flow Rate160 mL/min.
Shelf Life18 months from date of manufacture
Unit Size5 kits