Optical Adhesive Covers GPLE
Optical Adhesive Covers GPLE
Applied Biosystems™

Optical Adhesive Covers GPLE

The Applied Biosystems Optical Adhesive Cover GPLE is a pressure sensitive film that reduces well-to-well contamination and prevents sample evaporationRead more
Have Questions?
Catalog NumberQuantity
A4976725 covers
Catalog number A49767
Price (USD)
113.65
Online Exclusive
119.00
Save 5.35 (4%)
Each
-
Add to cart
Quantity:
25 covers
Price (USD)
113.65
Online Exclusive
119.00
Save 5.35 (4%)
Each
Add to cart
The Applied Biosystems Optical Adhesive Cover GPLE is a pressure sensitive film that reduces well-to-well contamination and prevents sample evaporation when applied to a microplate. It is optically clear for use with real-time PCR applications. Each lot of these general purpose laboratory equipment (GPLE) covers is manufactured in an ISO registered facility making them ideal for use with diagnostic procedures.

Features of the Optical Adhesive Covers GPLE include:
• Reduce contamination as pressure-sensitive adhesive provides a tight seal over every well
• Optically clear for use with real-time PCR applications
• Labeled ‘For Laboratory Use’
• Designed to fit MicroAmp 96-well and 384-well plates

The Optical Adhesive Covers are easy to use. Simply peel the backing away from the adhesive film and place on the microplate. Use in combination with the MicroAmp Adhesive Film Applicator to apply pressure and form a tight seal between the microplate and the adhesive film. This will reduce the possibility of cross-contamination between sample wells and prevent sample evaporation for consistent real-time PCR data.

For Laboratory Use
Specifications
Certifications/ComplianceGPLE
For Use With (Application)PCR, qPCR
Includes25 covers
Product TypeFilm, Seal, or Applicator
Quantity25 covers
Shipping ConditionRoom temperature
AdhesivePressure Sensitive
PierceableNo
Plate Compatibility96/384-well Plate
SterilityNon-sterile
Unit SizeEach
Contents & Storage
Store at room temperature and avoid direct exposure to sunlight.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link