Gibco™

Viral-Plex™ Complexation Buffer

Catalog number: A4983901
Have Questions?
Gibco™

Viral-Plex™ Complexation Buffer

Catalog number: A4983901
Have Questions?
Catalog Number
A4983901
Unit Size
100 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A4983901100 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Gibco Viral-Plex Complexation Buffer is a chemically defined, protein-free, phenol red-free buffer for complexing plasmid DNA with transfection reagent for standard transfection applications.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Line
HEK293
Culture Type
Suspension Cell Culture
Description
Serum-free, animal origin-free, chemically defined, protein free, phenol red free
Cell Type
Established Cell Lines
Format
Bottle
Sample Type
Plasmid DNA
Transfection Technique
Lipid-based Transfection
Expression System
Mammalian
For Use With (Equipment)
AAV-MAX System
For Use With (Application)
AAV Production
Includes
100 mL Gibco Viral-Plex Complexation Buffer
Product Line
AAV-MAX
Product Type
AAV production transfection buffer
Quantity
100 mL
Reagent Type
Transfection buffer
Shelf Life
12 months from date of manufacture
Shipping Condition
Wet Ice
Sterility
Sterile

Contents & Storage

100mL bottle, store at 2°C to 8°C, protected from light. Do not freeze.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text