Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Description | Qualyfast Legionella Quantification Standards 8 tests |
Detection Method | qPCR |
Form | Lyophilized |
Format | Tube |
GC-Rich PCR Performance | Low |
PCR Method | qPCR |
Reaction Speed | Fast |
No. of Tests | 8 Tests |
Product Type | Legionella PCR and DNA Extraction Kit |
Sufficient For | 8 Reactions |
For Use With (Application) | Pathogen Detection, DNA Extraction |
No. of Reactions | 8 Reactions |
Unit Size | Each |
Description | Qualyfast Legionella PCR Assay |
Detection Method | Primer-probe |
Form | Lyophilized |
Format | Tube |
GC-Rich PCR Performance | Low |
PCR Method | qPCR |
Reaction Speed | Fast |
For Use With (Equipment) | Applied Biosystems 7500 Fast Food Safety Real-Time PCR instrument and Applied Biosystems QuantStudio 5 Food Safety Real Time PCR System |
No. of Tests | 24 Tests |
Product Type | Legionella PCR and DNA Extraction Kit |
Sufficient For | 8 Reactions |
For Use With (Application) | Pathogen Detection, DNA Extraction |
No. of Reactions | 8 Reactions |
Unit Size | Each |
Description | Qualyfast Legionella PCR Assay |
Detection Method | Primer-probe |
Form | Lyophilized |
Format | Tube |
GC-Rich PCR Performance | Low |
PCR Method | qPCR |
Reaction Speed | Fast |
For Use With (Equipment) | Applied Biosystems 7500 Fast Food Safety Real-Time PCR instrument and Applied Biosystems QuantStudio 5 Food Safety Real Time PCR System |
No. of Tests | 48 Tests |
Product Type | Legionella PCR and DNA Extraction Kit |
Sufficient For | 8 Reactions |
For Use With (Application) | Pathogen Detection, DNA Extraction |
No. of Reactions | 8 Reactions |
Unit Size | Each |
Description | Qualyfast Legionella PCR Assay |
Detection Method | Primer-probe |
Form | Lyophilized |
Format | Tube |
GC-Rich PCR Performance | Low |
PCR Method | qPCR |
Reaction Speed | Fast |
For Use With (Equipment) | Applied Biosystems 7500 Fast Food Safety Real-Time PCR instrument and Applied Biosystems QuantStudio 5 Food Safety Real Time PCR System |
No. of Tests | 96 Tests |
Product Type | Legionella PCR and DNA Extraction Kit |
Sufficient For | 8 Reactions |
For Use With (Application) | Pathogen Detection, DNA Extraction |
No. of Reactions | 8 Reactions |
Unit Size | Each |
Description | Qualyfast DNA Extraction Kit I – Clear Water |
Detection Method | qPCR |
Form | Lyophilized |
Format | Tube |
GC-Rich PCR Performance | Low |
PCR Method | qPCR |
Reaction Speed | Fast |
Product Type | Legionella PCR and DNA Extraction Kit |
Sufficient For | 8 Reactions |
For Use With (Application) | Standard PCR, DNA Extraction |
No. of Reactions | 24 Reactions |
Unit Size | Each |
Description | Qualyfast DNA Extraction Kit I – Clear Water |
Detection Method | qPCR |
Form | Lyophilized |
Format | Tube |
GC-Rich PCR Performance | Low |
PCR Method | qPCR |
Reaction Speed | Fast |
Product Type | Legionella PCR and DNA Extraction Kit |
Sufficient For | 8 Reactions |
For Use With (Application) | Standard PCR, DNA Extraction |
No. of Reactions | 48 Reactions |
Unit Size | Each |
Description | Qualyfast DNA Extraction Kit II – Dirty Water |
Detection Method | qPCR |
Form | Lyophilized |
Format | Tube |
GC-Rich PCR Performance | Low |
PCR Method | qPCR |
Reaction Speed | Fast |
Product Type | Legionella PCR and DNA Extraction Kit |
Sufficient For | 8 Reactions |
For Use With (Application) | Standard PCR, DNA Extraction |
No. of Reactions | 48 Reactions |
Unit Size | Each |
Description | Qualyfast DNA Extraction Kit I – Clear Water |
Detection Method | qPCR |
Form | Lyophilized |
Format | Tube |
GC-Rich PCR Performance | Low |
PCR Method | qPCR |
Reaction Speed | Fast |
Product Type | Legionella PCR and DNA Extraction Kit |
Sufficient For | 8 Reactions |
For Use With (Application) | Standard PCR, DNA Extraction |
No. of Reactions | 96 Reactions |
Unit Size | Each |
Description | Qualyfast DNA Extraction Kit II – Dirty Water |
Detection Method | qPCR |
Form | Lyophilized |
Format | Tube |
GC-Rich PCR Performance | Low |
PCR Method | qPCR |
Reaction Speed | Fast |
Product Type | Legionella PCR and DNA Extraction Kit |
Sufficient For | 8 Reactions |
For Use With (Application) | Standard PCR, DNA Extraction |
No. of Reactions | 96 Reactions |
Unit Size | Each |
Description | Qualyfast free DNA removal |
Detection Method | qPCR |
Form | Lyophilized |
Format | Tube |
GC-Rich PCR Performance | Low |
PCR Method | qPCR |
Reaction Speed | Fast |
Product Type | Legionella PCR and DNA Extraction Kit |
Sufficient For | 200 Reactions |
For Use With (Application) | Food & Beverage |
No. of Reactions | 200 Reactions |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | No. of Reactions | No. of Tests | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
A501013 | Each | Qualyfast Legionella Quantification Standards 8 tests | 8 Reactions | 8 Tests | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A101013 | Each | Qualyfast Legionella PCR Assay | 8 Reactions | 24 Tests | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A201013 | Each | Qualyfast Legionella PCR Assay | 8 Reactions | 48 Tests | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A301013 | Each | Qualyfast Legionella PCR Assay | 8 Reactions | 96 Tests | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
D101001 | Each | Qualyfast DNA Extraction Kit I – Clear Water | 24 Reactions | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
D201001 | Each | Qualyfast DNA Extraction Kit I – Clear Water | 48 Reactions | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
D211001 | Each | Qualyfast DNA Extraction Kit II – Dirty Water | 48 Reactions | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
D301001 | Each | Qualyfast DNA Extraction Kit I – Clear Water | 96 Reactions | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
D311001 | Each | Qualyfast DNA Extraction Kit II – Dirty Water | 96 Reactions | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
D401009 | Each | Qualyfast free DNA removal | 200 Reactions | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Minimizes false negatives
Increases accuracy
Easy-to-use
Optimized workflow
The qualyfast™ Legionella PCR Assay contains:
The qualyfast™ DNA Extraction Kit I is designed for use with clear water samples, as acquired from water distribution systems, drinking water and other sources that contain minimal suspended particles. The kit contains:
The qualyfast™ DNA Extraction Kit II is designed for use with dirty water, biofilm and sediment samples. The kit contains:
The qualyfast™ Free DNA Inactivator Kit contains:
The qualyfast™ Legionella Quantification Standards Kit contains:
The qualyfast™ Legionella workflow is compatible with the Applied Biosystems™ 7500 Fast Food Safety Real-Time PCR System or Applied Biosystems™ QuantStudio™ 5 Food Safety Real-Time PCR System.