Sterilized Sample Ports for CTS™ Rotea™ Single-Use Kit
Product Image
Sterilized Sample Ports for CTS™ Rotea™ Single-Use Kit
The Gibco CTS Rotea Single-Use Kit is part of the Gibco CTS Rotea Counterflow Centrifugation System. Each kit can performRead more
Promo ImagePromotion
Promo code:RPUZZ25
Stock up on essentials to piece your discovery together
Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzleLearn More
Have Questions?
Catalog NumberQuantity
A50111Promo Image10 ports
Catalog number A50111
Price (USD)
487.00
Each
In stock
Add to cart
Quantity:
10 ports
Recurring order eligible. Learn more »
Price (USD)
487.00
Each
Add to cart
Ask our AI about this Product
The Gibco CTS Rotea Single-Use Kit is part of the Gibco CTS Rotea Counterflow Centrifugation System. Each kit can perform a wide range of cell processing steps, making it ideal for wash & concentration, separation, isolation, and more. The single-use kit provides flexibility for process development and easily supports commercial-scale manufacturing. Each kit is compatible with sterile luer connectors or thermal welding.
These Sterilized Sample Ports for use with the kit enable collection of product samples during protocol execution, for process development. The sample port assembly consists of a needleless port joined with short lengths of PVC tube. The PVC tube is sealed to enable thermal welding. The sample port assemblies and PVC tubing are gamma-irradiated and individually packaged.
Specifications
Quantity10 ports
Capacity10 Pk.
Product LineCTS Rotea
RefrigeratedNo
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Customers who viewed this item also viewed
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Frequently asked questions (FAQs)
The Rotea Protocol Builder Software, which includes the Rotea Process Model and Simulator, is available for download from the Rotea website. Common cell types are already pre-loaded into the software, and custom cells can also be created. The CTS Rotea System can separate cells that have at least 4 µm difference in diameter. Thermo Fisher Scientific recommends that customers use protocol optimization and workflow validation testing to meet their process needs. Please visit our webpage (www.thermofisher.com/rotea) to see if the CTS Rotea System will work for your cell type, or submit a request for more information using our demo form (https://www.thermofisher.com/us/en/home/global/forms/life-science/rotea-counterflow-centrifugation-system-cell-therapy.html) to get connected with a Business Development Manager.
Thermo Fisher Scientific strives to provide a catalog of tools to support our diverse customers' needs. If magnetic bead separation using the CTS Rotea System is of interest, Thermo Fisher Scientific recommends that customers use protocol optimization and workflow validation testing to meet their process needs.
Ficoll medium has many shortcomings and often does not scale well for cGMP manufacturing. Rotea PBMC isolation protocol takes less time and has been demonstrated to be capable of providing higher cell recovery compared to Ficoll medium. Thermo Fisher Scientific recommends that customers use protocol optimization and workflow validation testing to meet their process needs.
The CTS Rotea single-use kits are manufactured with DEHP-free PVC. Extractables and Leechables testing was conducted with 20% DMSO for up to 15 min. The CTS Rotea System has not been validated for fill-finish applications. Thermo Fisher Scientific recommends that customers use protocol optimization and workflow validation testing to meet their process needs.
The CTS Rotea System has been demonstrated to be capable of washing cells from cryoprotectant into media and the reverse. The CTS Rotea System has not been validated for fill-finish applications. Thermo Fisher Scientific recommends that customers use protocol optimization and workflow validation testing to meet their process needs
Share catalog number, name or link
1x1 image pixel for data collection