Gibco™

CTS Xenon SingleShot Electroporation Chamber

Catalog number: A50305
Have Questions?
Gibco™

CTS Xenon SingleShot Electroporation Chamber

Catalog number: A50305
Have Questions?
Catalog Number
A50305
Unit Size
6 chambers
Price (USD)
Catalog NumberUnit SizePrice (USD)
A503056 chambersRequest A Quote
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Gibco CTS Xenon SingleShot Electroporation Chamber is for use with the Xenon Electroporation for cell therapy research. This single-use electroporation chamber is made of Tritan™ MX711 co-polyester with gold-plated electrodes and sterilized by gamma irradiation. This chamber can process 2 × 107 to 1 × 108 T cells in a 1-mL sample volume and is offered in a pack of six individually packaged chambers.

Promising electroporation conditions that have been optimized in the Neon Transfection System can be scaled to the Xenon Electroporation System without sacrificing performance.

View an interactive 3D tour of the CTS Xenon instrument ›

Specifications

Disposable
Single-use
Capacity (Metric)
1 mL
Material
Tritan MX711 Copolyester
Quantity
6 chambers
For Use With (Equipment)
Gibco CTS Xenon Electroporation System
For Use With (Application)
Cell and Gene Therapy Research, Development, and Manufacturing
Sterilization Method
Gamma Irradiation
Shelf Life
24 Months from Date of Manufacture
Manufacturing Quality
ISO 13485

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text