Gibco™

CTS™ B-27™ Supplement XenoFree (50X)

Catalog number: A5047501
Have Questions?
Gibco™

CTS™ B-27™ Supplement XenoFree (50X)

Catalog number: A5047501
Have Questions?
Catalog Number
A5047501
Unit Size
10 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A504750110 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
CTS B-27 Supplement XenoFree is based on the classic serum-free B-27 supplement and designed to help meet regulatory requirements for cell therapy manufacturing. It supports the growth and differentiation of a variety of cell types derived from pluripotent stem cells (PSCs). Example applications include induction of human neural stem cells (hNSCs), expansion of hNSCs, differentiation of hNSCs, and maintenance of mature differentiated neurons in culture.

CTS quality
Gibco CTS products are manufactured at a site that uses methods and controls that conform with cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing sites are ISO 13485– and ISO 9001–certified.

Accompanied by documentation to support your regulatory filing
• Access to our Drug Master File (DMF) or Regulatory Support File (RSF)
• Traceability documentation—Certificate of Origin, Certificate of Analysis, and Safety Data Sheet

Reduced risk from animal-origin materials and human-derived components
CTS B-27 Supplement XenoFree is formulated with only AOF recombinant components and human-derived components. To help reduce viral clearance risks, the use of human-derived components has been minimized and human-derived components are limited to materials that help meet regulatory standards for donor qualification, viral testing, and viral inactivation.

Specifications

Classification
Serum-free, Xeno-free
Concentration
50X
Culture Type
Cells derived from Pluripotent Stem Cells (PSCs)
Form
Liquid
Product Type
Supplement
Shelf Life
12 months from date of manufacture
Sterility
Sterile-filtered
Manufacturing Quality
ISO 13485, MDSAP, FDA-registered, 21 CFR 820
Quantity
10 mL
Shipping Condition
Dry Ice

Contents & Storage

Store at -5°C to -20°C and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text