PrepSEQ™ Nucleic Acid Sample Preparation Kit
PrepSEQ™ Nucleic Acid Sample Preparation Kit
Applied Biosystems™

PrepSEQ™ Nucleic Acid Sample Preparation Kit

The Applied Biosystems PrepSEQ Nucleic Acid Sample Preparation Kit is optimized for highly efficient, quantitative, and consistent recovery of DNARead more
Have Questions?
Catalog NumberQuantity
A50485100 preps
Catalog number A50485
Price (USD)
1,206.00
Each
Add to cart
Quantity:
100 preps
Price (USD)
1,206.00
Each
Add to cart

The Applied Biosystems PrepSEQ Nucleic Acid Sample Preparation Kit is optimized for highly efficient, quantitative, and consistent recovery of DNA and RNA from diverse samples types, including cell culture, raw material (e.g., serum), and in-process downstream purification samples such as column purification or bulk drug substance (BDS) samples.

  • Time-saving, cost-effective DNA and RNA extraction for diverse sample types
  • Reproducible recovery of DNA and RNA from complex samples
  • Manual and high-throughput automated workflow on KingFisher Flex instrument, isolating up to 24 samples (in triplicate) in 5 hours

Easy to use

The PrepSEQ Nucleic Acid Sample Preparation Kit provides a high-throughput automated solution in combination with the Thermo Scientific KingFisher Flex instrument for low hands-on extraction of DNA and RNA.

Consistent performance

For accurate quantitation of DNA and RNA, extraction consistency must be unaffected by complex matrices. Our advanced magnetic bead-based technology provides highly consistent extraction performance to support reliable quantitation.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Impurity Testing
For Use With (Equipment)KingFisher Flex
High-throughput CompatibilityNot High-throughput Compatible (Manual), Automated Protocols
No. of Reactions100 Reactions
Product LinePrepSEQ™
Quantity100 preps
Sample TypeBiopharmaceutical Production Samples
Sample Volume100 μL
TargetGenomic D, R, Viral R
Test Time2 hr.
Isolation TechnologyMagnetic Bead
Unit SizeEach
Please enter sku to load default questions

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link