Gibco™

AAV-MAX Lysis Buffer

Catalog number: A50520
Have Questions?
Gibco™

AAV-MAX Lysis Buffer

Catalog number: A50520
Have Questions?
Catalog Number
A50520
Unit Size
100 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A50520100 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Gibco AAV-MAX Lysis Buffer is a ready-to-use cell lysis reagent for the extraction of adeno-associated virus (AAV) particles from producer HEK293 cells. Cell lysis is detergent-based, eliminating the need for mechanical cell disruption, providing a milder and easier alternative when isolating AAV particles from cell cultures. The buffer is supplied as a 10X solution that can be directly added to HEK293 AAV production cultures to induce cell lysis.

Gibco AAV-MAX Lysis Buffer is a Polysorbate 20-based formulation and does not contain Triton™ X-100.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Concentration
10X
Product Type
Cell Lysis Buffer
Shelf Life
12 months from date of manufacture
Shipping Condition
Wet Ice
Form
Liquid
Product Line
AAV-MAX
For Use With (Application)
AAV Production
Purification Target
AAV
Sufficient For
1 L of Culture
Quantity
100 mL
Composition
Polysorbate 20 based
pH
6–8

Contents & Storage

100mL Bottle, Store at 2°C to 8°C

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text