Pierce™ High Capacity Ni-IMAC Resin, EDTA compatible
Pierce™ High Capacity Ni-IMAC Resin, EDTA compatible
Thermo Scientific™

Pierce™ High Capacity Ni-IMAC Resin, EDTA compatible

Pierce high-capacity EDTA-compatible Ni-IMAC resin is designed for the purification of over-expressed His-tagged proteins from cell culture media. The matrixRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A50586250 mL
A5058410 mL
A5058550 mL
A50587500 mL
Catalog number A50586
Price (USD)
2,813.65
Online Exclusive
3,130.00
Save 316.35 (10%)
Each
-
Add to cart
Quantity:
250 mL
Price (USD)
2,813.65
Online Exclusive
3,130.00
Save 316.35 (10%)
Each
Add to cart
Pierce high-capacity EDTA-compatible Ni-IMAC resin is designed for the purification of over-expressed His-tagged proteins from cell culture media. The matrix is a 6% crosslinked agarose with optimal properties for batch, gravity column, and research/pilot scale FPLC column purifications. The resin has high binding capacity (up to 80 mg of protein per mL of resin) and can tolerate the high EDTA and reducing agent concentrations used to inhibit metalloproteases and stabilize sensitive intracellular proteins.

Features of the high-capacity EDTA-compatible Ni-IMAC resin include:
• Chelator stable—use up to 20 mM EDTA and DTT with no loss in performance
• Compatible with purification of overexpressed, secreted proteins in cell culture media (e.g., Expi293, ExpiCHO, and ExpiSf expression systems)
• High binding capacity—>80 mg/mL binding capacity for His-tagged recombinant proteins
• Reusable—can be regenerated up to 10 times using the CIP protocol

EDTA and reducing agents such as DTT and TCEP adversely affect the performance of Ni-IMAC supports by stripping off the metal. Because of its chelator stability characteristic (EDTA tolerant), Pierce high-capacity EDTA-compatible Ni-IMAC resin is especially suited for purifying expressed His-tagged proteins that are secreted into cell culture media or for purifying intracellular His-tagged proteins that need the presence of EDTA to maintain stability and function. In both cases, large quantities of samples can be purified without the Ni metal stripping off of the support, thereby providing consistent high capacity yields.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Binding PropertyChelator Stable
Column TypeBottle
Matrix6% crosslinked agarose
Quantity250 mL
Shipping ConditionWet ice
Product LinePierce™
TypeResin
Unit SizeEach
Contents & Storage
250 mL settled resin, supplied as 500 mL 50% slurry. Store at 2–8°C.

Frequently asked questions (FAQs)

With Pierce High Capacity Ni-IMAC Resin, EDTA compatible, what residence times are adequate for FPLC purification?

Optimized residence times will differ by target, but the standard range is 2-10 mins.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

How is Pierce High Capacity Ni-IMAC Resin, EDTA compatible different from standard Ni-NTA resin?

Pierce High Capacity Ni-IMAC Resin, EDTA compatible doesn't leach and discolor in the presence of reducing agent, EDTA, or cell culture supernatant, thereby providing exceptionally high protein yield.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

After regeneration of Pierce High Capacity Ni-IMAC Resin, EDTA compatible, is there a specific buffer the resin should be stored in?

The recommended storage buffer for the column and resin is 20% ethanol, 10 mM sodium acetate, pH 6.50. Please refer to the product manual (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0024862_PierceHighCapNiIMACResin_UG.pdf) for details.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

With Pierce High Capacity Ni-IMAC Resin, EDTA compatible, how do I regenerate the resin?

Pierce High Capacity Ni-IMAC Resin, EDTA compatible is alkali-stable. The resin can be exposed to NaOH, washed with water, neutralized with PBS, washed with water, and put back into storage buffer. Please refer to the product manual (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0024862_PierceHighCapNiIMACResin_UG.pdf) for details.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

How many uses/regenerations are possible with Pierce High Capacity Ni-IMAC Resin, EDTA compatible?

Up to 10 uses are possible with Pierce High Capacity Ni-IMAC Resin, EDTA compatible.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link