Thermo Scientific™

Pierce™ High Capacity Ni-IMAC MagBeads, EDTA compatible

Catalog number: A50588
Have Questions?
Thermo Scientific™

Pierce™ High Capacity Ni-IMAC MagBeads, EDTA compatible

Catalog number: A50588
Have Questions?
Catalog Number
A50588
Unit Size
1 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
5 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
25 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
100 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A505881 mL
-
A505895 mL
-
A5059025 mL
-
A50591100 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Pierce high-capacity EDTA-compatible Ni-IMAC MagBeads are designed for purification of over-expressed His-tagged proteins from cell culture media. The beads have high binding capacity (up to 80 mg of protein per mL of settled beads) and can tolerate the high EDTA and reducing agent concentrations that are used to inhibit metalloproteases and stabilize sensitive intracellular proteins.

Features of the high-capacity EDTA-compatible Ni-IMAC MagBeads include:
• Chelator stable—stable in buffer containing 20 mM EDTA and DTT
• Compatible with purification of overexpressed, secreted proteins in cell culture media (e.g., Expi293, ExpiCHO, and ExpiSf expression systems)
• Exceptional high protein binding capacity—up to 80 mg/mL of settled beads
• High-performance—non-aggregating, superparamagnetic microparticles provide exceptional performance with manual and automated HTS applications (e.g., Thermo Scientific KingFisher instruments)

Pierce high-capacity EDTA-compatible Ni-IMAC MagBeads consist of crosslinked agarose beads embedded with magnetite. The density of the ligand on the magnetic agarose bead results in a binding capacity similar to or better than traditional agarose resins with the added benefits of having magnetic properties and being EDTA compatible. The MagBeads are a valuable tool for small-scale purification of multiple His-tagged proteins and for scouting expression and purification conditions to be used in larger-scale purifications.

Pierce high-capacity EDTA-compatible Ni-IMAC MagBeads are optimized for use with high-throughput magnetic platforms, such as the KingFisher 96 and KingFisher Flex magnetic particle processors, but the beads also compatible with simple benchtop purification applications using an appropriate magnetic stand. Automated instruments, however, are especially useful for higher throughput purification and screening of purification conditions.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Binding Capacity
Up to 80 mg of 6 xHis-tagged protein / mL of settled magnetic beads
Binding Property
Chelator Stable
Description
EDTA compatible Ni-IMAC magnetic beads for His-tag protein purification
Matrix
Magnetic Agarose
pH Range
2 to 14 pH stability
Shipping Condition
Wet Ice
Particle Size
30 μm
Form
Liquid Suspension
Product Line
Pierce™
Quantity
1 mL
Type
Magnetic Affinity Bead
Column Type
Bottle

Contents & Storage

1 mL of 25% suspension. Store at 2–8°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text