Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Green Features | Less waste |
Assay | Qubit Protein Broad Range Assay |
Detection Method | Fluorescence |
Product Line | Qubit™ |
Product Type | Protein BR Assay Kit |
Sufficient For | 500 Reactions |
For Use With (Equipment) | Qubit 4 Fluorometer |
Quantitation Range | 0.1 to 20 mg/mL |
Quantity | 400 Assays |
Shipping Condition | Wet Ice |
Unit Size | 500 assays |
Assay | Qubit Protein Assay |
Detection Method | Fluorescence |
Product Line | Qubit™ |
Product Type | Protein Quantification Assay |
Sufficient For | 100 Reactions |
For Use With (Equipment) | Qubit Fluorometer, Microplate Reader |
Quantitation Range | 12.5 μg/mL to 5 mg/mL |
Quantity | 100 Assays |
Shipping Condition | Room Temperature |
Unit Size | 100 assays |
Assay | Qubit Protein Assay |
Detection Method | Fluorescence |
Product Line | Qubit™ |
Product Type | Protein Quantification Assay |
Sufficient For | 500 Reactions |
For Use With (Equipment) | Qubit Fluorometer, Microplate Reader |
Quantitation Range | 12.5 μg/mL to 5 mg/mL |
Quantity | 500 Assays |
Shipping Condition | Room Temperature |
Unit Size | 500 assays |
Green Features | Less waste |
Assay | Qubit Protein Broad Range Assay |
Detection Method | Fluorescence |
Product Line | Qubit™ |
Product Type | Protein BR Assay Kit |
Sufficient For | 100 Reactions |
For Use With (Equipment) | Qubit 4 Fluorometer |
Quantitation Range | 0.1 to 20 mg/mL |
Quantity | 100 Assays |
Shipping Condition | Wet Ice |
Unit Size | 100 assays |
Catalog Number | Specifications | Unit Size | Assay | Quantitation Range | Sufficient For | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
A50669 | 500 assays | Qubit Protein Broad Range Assay | 0.1 to 20 mg/mL | 500 Reactions | 298.65 Online price 307.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Q33211 | 100 assays | Qubit Protein Assay | 12.5 μg/mL to 5 mg/mL | 100 Reactions | 128.65 Online price 136.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Q33212 | 500 assays | Qubit Protein Assay | 12.5 μg/mL to 5 mg/mL | 500 Reactions | 387.65 Online price 413.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A50668 | 100 assays | Qubit Protein Broad Range Assay | 0.1 to 20 mg/mL | 100 Reactions | 149.65 Online price 154.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|