Multiskan SkyHigh Microplate Spectrophotometer is a UV/Vis microplate reader designed to be convenient and easy to use for any photometric absorbtion or turbidimetric research applications and especially DNA, RNA and protein analysis. This microplate reader is ideal for multi-user environments where a variety of endpoint, kinetic and spectral assays are performed. Multiskan SkyHigh is available in different configurations. Touchscreen models offer the flexibility to use the system as stand-alone, or together with Thermo Scientific™ SkanIt Software for PC control. A model with a cuvette reading option is also available. For convenient data handling, Multiskan SkyHigh has an option to access to Thermo Fisher Connect and Microsoft OneDrive cloud-based tools.
Catalog Number
A51119500C
Unit Size
Each
Description
Multiskan SkyHigh
dimReadingOptions
Price (USD)
15,670.00
Availability
-
Full specifications
Quantity
Cuvettes
No
Incubator Temperature
Ambient +2° to 45°C
Light Source
Xenon Flash Lamp
Measurement Speed
96 wells: 6 sec., 384 wells: 10 sec.
Plate Type
6, 48, 96, and 384-well Plates, μDrop Plates
Product Line
Multiskan
Shaking
Linear
Software Type
SkanIt Software, Open License Software, Multi-Instrument Software
Spectral Scanning Speed
10 sec. from 200 to 1000 nm
Wavelength Selection
Monochromator
Accuracy (Photometric)
1.0% + 0.003 Abs (0 to 2.0 Abs), 2.0% (2.0 to 2.5 Abs) at 450 nm
Bandwidth
<2.5 nm
Depth (English)
17.5 in.
Depth (Metric)
44.5 cm
Description
Multiskan SkyHigh
Height (English)
10.4 in.
Height (Metric)
26.5 cm
Linearity
0 to 2.5 Abs, 2% at 450 nm
Resolution
1 nm
Software Language Versions
English, German, French, Spanish, Italian, Portuguese, Russian, Chinese, Japanese
Multiskan SkyHigh with touchscreen, cuvette and μDrop Duo Plate
22,820.00
-
Cuvettes
Yes
Incubator Temperature
Ambient +2° to 45°C
Interface
Touchscreen
Light Source
Xenon Flash Lamp
Measurement Speed
96 wells: 6 sec., 384 wells: 10 sec.
Plate Type
6, 48, 96, and 384-well Plates, μDrop Plates
Product Line
Multiskan
Shaking
Linear
Software Type
SkanIt Software, Open License Software, Multi-Instrument Software
Spectral Scanning Speed
10 sec. from 200 to 1000 nm
Wavelength Selection
Monochromator
Accuracy (Photometric)
1.0% + 0.003 Abs (0 to 2.0 Abs), 2.0% (2.0 to 2.5 Abs) at 450 nm
Bandwidth
<2.5 nm
Depth (English)
17.5 in.
Depth (Metric)
44.5 cm
Description
Multiskan SkyHigh with touchscreen, cuvette and μDrop Duo Plate
Height (English)
10.4 in.
Height (Metric)
26.5 cm
Includes
µDrop Duo plate
Linearity
0 to 2.5 Abs, 2% at 450 nm
Resolution
1 nm
Software Language Versions
English, German, French, Spanish, Italian, Portuguese, Russian, Chinese, Japanese
Wavelength Range
200 to 1000 nm
Weight (English)
23.8 lb.
Weight (Metric)
10.8 kg
Width (English)
11.6 in.
Width (Metric)
29.5 cm
Unit Size
Each
Showing 5 of 5
Monochromator-based optical system for free selection of wavelengths from 200 nm to 1000 nm
Compatible with 6-48, 96- and 384-well microplates or cuvettes with maximum plate height of 19.5 mm including possible lid
Reads μDrop™ and μDrop Duo Plates for micro-volume DNA/RNA and protein analysis
Separate optimized measurement modes for absorption and turbidimetric measurements
Allows kinetic, spectral, and endpoint measurements for a variety of applications
Fast operation: full spectrum of a well is done in less than 10 seconds; typical DNA spectrum can be measured in less than three seconds and a full 96-well microplate is read with one wavelength in 6 seconds
Performs onboard shaking and incubation for temperature-critical or cell-based assays
Touchscreen models have an easy-to-use interface for stand-alone use, and includes ready-made protocols for UV based nucleic acid and protein quantification as well as colorimetric protein quantification
Thermo Fisher Connect or Microsoft OneDrive cloud-based tools allow you to securely store, access, share, and manage your photometric data remotely (touchscreen models)
Multiple languages available; choose between English, German, French, Spanish, Italian, Portuguese, Russian, Chinese, and Japanese (touchscreen models)
Setting up of more demanding assays is easy via the powerful and intuitive SkanIt™ Software
Multi-instrument software platform compatible with all Thermo Scientific microplate readers
Open license software for unlimited installation to multiple computers
Research Edition for life sciences; Drug Discovery Edition with features for compliance with FDA 21 CFR Part 11
User-interface languages available: English, French, German, Spanish, Portuguese, Italian, Russian, Japanese, and Chinese
Searchable SkanIt Cloud Library with ready-to-use protocols including calculations and true application example data
Measurement sessions are easy to setup and navigate
Intuitive Virtual Pipetting Tool assists in defining the plate layout