Informational:Discover the future of Gibco Media selection with our upcoming updated experience. View details >
Gibco™

CTS™ Viral Production Medium

Catalog number: A5144003
Have Questions?
Gibco™

CTS™ Viral Production Medium

Catalog number: A5144003
Have Questions?
Catalog Number
A5144003
Unit Size
10 L
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1 L
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
6 x 1 L
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
20 L
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
100 L
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A514400310 L
-
A51440011 L
-
A51440026 x 1 L
-
A541600120 L
-
A5416002100 L
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

Gibco CTS Viral Production Medium is a chemically defined, serum-free, protein-free, animal-origin-free, and phenol-red-free medium that is part of the CTS AAV-MAX Helper-Free AAV Production System. CTS Viral Production Medium is specifically developed for growth and transfection of CTS Viral Production Cells 2.0 (suspension-adapted HEK293F-derived cells) for adeno-associated virus (AAV) production.

Features of CTS Viral Production Medium include:

  • Supports growth and transfection of CTS Viral Production Cells 2.0 from shake flask to bioreactor scales
  • Enables production of high AAV titers, greater than 5 x 1010 vg/mL (unpurified)
  • Formulated with GlutaMAX Supplement, so does not require further supplementation. With the exception of containing GlutaMAX Supplement, the formulation of this product is identical to research-grade Viral Production Medium.
  • Manufactured in conformity with cGMP for medical devices (21 CFR Part 820) and follows USP <1043> and European Pharmacopoeia (Ph. Eur.) 5.2.12 recommendations. Drug Master File and Regulatory Support File available upon request

The CTS AAV-MAX Helper-Free AAV Production System is a scalable, high-titer platform for suspension AAV production with research-grade and GMP versions to enable seamless transition from research to commercial manufacturing. Other components of the CTS AAV-MAX system include CTS Viral Production Cells 2.0, CTS AAV-MAX Transfection Kit, CTS Viral-Plex Complexation Buffer, and Viral Production Medium, AGT.

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.

Specifications

Cell Line
Viral Production Cells 2.0
Cell Type
HEK293
Classification
Animal Origin-free
Form
Liquid
Product Type
AAV Production Medium
Shelf Life
12 months from date of manufacture
Sterility
Sterile-filtered
Format
BPC
For Use With (Application)
AAV Production
Quantity
10 L
Shipping Condition
Ambient

Contents & Storage

10 L bag. Store at 2°C to 8°C and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text