Gibco™

Viral Production Medium, AGT™

Catalog number: A5147103
Have Questions?
Gibco™

Viral Production Medium, AGT™

Catalog number: A5147103
Have Questions?
Catalog Number
A5147103
Unit Size
50 L
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1 L
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
10 L
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
100 L
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A514710350 L
-
A51471011 L
-
A514710210 L
-
A5147104100 L
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Gibco Viral Production Medium, AGT, is the dry-format version of CTS Viral Production Medium, a chemically defined, serum-free, protein-free, animal origin-free, phenol red-free medium that is part of the CTS AAV-MAX Helper-Free AAV Production System for high-titer adeno-associated virus (AAV) production.

Features of Viral Production Medium, AGT, include:
• Supports growth and transfection of CTS Viral Production Cells 2.0 from shake flask to bioreactor scales
• Helps enable production of high AAV titers, greater than 5 x 1010 vg/mL (unpurified)
• Simplifies transfer to manufacturing scale due to Advanced Granulation Technology (AGT) enabling auto-pH and simplified dissolution
• Formulated with GlutaMAX Supplement, no further supplementation required
• Manufactured in conformity with cGMP for medical devices, 21 CFR, Part 820, and certified to ISO 13485. Drug Master File and Regulatory Support File available upon request

Advanced Granulation Technology
Advanced Granulation Technology (AGT) helps simplify medium preparation at large scale with fast rehydration, a just-add water protocol, and low dust generation, creating less pressure build-up during sterile filtration. For more information on AGT media preparation at large scale, see Hydration and scale-up of AGT medium in HyPerforma and imPULSE Single-Use Mixers (PDF).

Design your own media specific to your needs
For large-scale biomanufacturing processes, we offer specialized packaging specific to your needs. With Gibco cGMP Media Custom Services, you can put the manufacturing power and years of technical experience of our R&D and Bioproduction Application Specialists to work for you.

For research use or further manufacturing. Not for diagnostic use or direct administration into humans or animals.

Specifications

Cell Line
Viral Production Cells 2.0
Cell Type
HEK 293 cells
Concentration
25.6 g/L
Culture Type
Suspension Cell Culture
Expression System
Mammalian
Form
Powder (AGT)
Product Type
AAV Production Medium
Shelf Life
12 Months
Sterility
Non-sterile
With Additives
Glucose, GlutaMAX
Without Additives
No Phenol Red, No Antibiotics
For Use With (Application)
AAV Production
pH
6.9 to 7.5
Product Line
AAV-MAX
Reconstituted Volume
50 L

Contents & Storage

50 L Ropack Bucket, store at 2°C to 8°C protected from light

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text