Ion Torrent™

Oncomine™ IGH FR3(d)-J Assay, DNA

Catalog number: A51560
Have Questions?
Ion Torrent™

Oncomine™ IGH FR3(d)-J Assay, DNA

Catalog number: A51560
Have Questions?
Catalog Number
A51560
Unit Size
24 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A5156024 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Oncomine IGH FR3(d)-J Assay is a targeted next-generation sequencing (NGS) assay for detection of B-cell diversity to measure clonal expansion/contraction with accuracy and for detection of rare clones with high sensitivity. Primers have been specifically designed to target the distal region of FR3 to J of the immunoglobulin heavy chain of the B-cell receptor. Due to lower levels of somatic hypermutation observed in the distal region of FR3, there is an increase in primer-binding efficiency. When combined with Ion Reporter Software, data can be easily analyzed and reports run within a short period of time.

Key features of the Oncomine IGH FR3(d)-J Assay:
• Sequencing of FR3(d)-J region of IGH chain
• Unique primer design to distal FR3 region may reduce primer mismatches due to somatic hypermutation
• Detect rare clones at LOD of 10-6
• Optimized Oncomine informatics for easy, all-inclusive analysis and publication-ready data plots

The kit provides reagents for library construction and dNTPs. Ion AmpliSeq libraries can be generated from DNA extracted from FFPE tissue, fresh/frozen tissue, bone marrow, whole blood, peripheral blood leukocytes, PBMCs, and sorted B cells with as little as 50 ng of nucleic acid. The assay is optimized for sequencing on the Ion GeneStudio S5 System with the Ion 530, Ion 540, and Ion 550 chips to enable up to 24 samples per run.

When the primary test isn’t enough
Often when the desired results are not achieved with a primary test, an assay designed with primers directed to different regions of the V(D)J region can be used as a secondary or follow-up test to improve sensitivity of detection. Where needed, secondary testing of samples can be achieved using the Oncomine IGH FR3(d)-J Assay.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Control Template
dNTP mix
Control Type
Ion AmpliSeq Library Plus Kit
Sample Type
DNA
Sequencing Type
Genome and DNA Sequencing
For Use With (Equipment)
GeneStudio
For Use With (Application)
Sequencing
Libraries
targeted sequencing
No. of Reactions
24
Shipping Condition
Dry Ice

Contents & Storage

• Oncomine IGH FR3(d)-J Panel
• dNTP mix
• Ion AmpliSeq Library Plus Kit

All components stored at -10 to -30°

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text