Ion Torrent™

Oncomine™ Myeloid Research Assay

Catalog number: A51770
Have Questions?
Ion Torrent™

Oncomine™ Myeloid Research Assay

Catalog number: A51770
Have Questions?
Catalog Number
A51770
Unit Size
96 reactions
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
24 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A5177096 reactions
-
A3694024 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Ion Torrent Oncomine Myeloid Research Assay consists of three pools of AmpliSeq oligonucleotide primers and associated reagents to generate amplicon libraries for next-generation sequencing (NGS) on Ion Torrent platforms. The assay is designed to provide sensitive and comprehensive sample amplification of relevant DNA mutations and fusion transcripts associated with myeloid disorders in a single NGS run. The panel is comprised of 40 key DNA genes and a broad fusion panel of 29 driver genes to cover the most relevant targets in major myeloid disorders: acute myeloid leukemia (AML), myeloid dysplastic syndrome (MDS), myeloproliferative neoplasms (MPN), chronic myeloid leukemia (CML), chronic myelomonocytic leukemia (CMML), and juvenile myelomonocytic leukemia (JMML).

This version of the assay is for manual library preparation. For automated library preparation using the Ion Chef System, please see Cat. No. A36941.

Features include:
• Simple, robust workflow that encompasses mutations and fusions in two DNA and one RNA pool in a single NGS run
• Excellent coverage of challenging targets such as CEBPA and FLT3-ITDs
• Multiplex up to four samples on a single Ion 318 chip or twelve on an Ion 530 chip
• Compatible with as little as 10 ng/pool input DNA or RNA per library from blood or bone marrow samples
• Validated detection of somatic variants down to 5% allele frequency
• Simple and fast workflow produces targeted libraries in 3.5 hours typically
• For use on the Ion PGM and Ion S5 systems with manual or automated library preparation
• Compatible with the Oncomine Knowledgebase Reporter for variant annotation

The Oncomine Myeloid Research Assay is a comprehensive, targeted NGS assay designed to assist hematologists in the understanding of myeloid cancer. The assay is a complete kit facilitating the amplification of multiple gene targets from blood or bone marrow samples with as little as 10 ng of input DNA or RNA per pool. Leveraging the power of Ion AmpliSeq technology, this highly multiplexed NGS assay enables the generation of results from multiple samples in a single run. Designed for use on either the Ion PGM or Ion S5 sequencing systems, results are delivered in days rather than weeks. The assay is aligned with bioinformatics workflows within Torrent Suite and Ion Reporter analysis software that utilize optimized variant-calling parameters for SNV, InDel, and FLT3 large internal tandem duplication (ITD) detection. Variants can be annotated in the Oncomine Knowledgebase Reporter to generate a sample-specific report. Samples can be processed quickly and easily, and variants detected and identified confidently.

Specifications

Sample Type
DNA
Sequencing Type
Genome and DNA Sequencing
For Use With (Equipment)
Ion PGM™ System
Libraries
Targeted Sequencing Library
No. of Reactions
96
Product Line
Oncomine™
Shipping Condition
Dry Ice
Workflow Step
DNA Extraction

Contents & Storage

Store in freezer (-5 to -30°C).

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text