Informational:Inquire about OEM or Commercial Supply version of this product here.
Invitrogen™

SuperScript™ IV RT-LAMP Master Mix

Catalog number: A51801
Have Questions?
Invitrogen™

SuperScript™ IV RT-LAMP Master Mix

Catalog number: A51801
Have Questions?
Catalog Number
A51801
Unit Size
100 reactions
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
400 reactions
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1000 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A51801100 reactions
-
A51802400 reactions
-
A518031000 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

Invitrogen SuperScript IV RT-LAMP Master Mix is a ready-to-use solution optimized for fast, sensitive, and simple RNA target detection in loop-mediated isothermal amplification (LAMP) reactions. With this convenient master mix formulation, you can perform isothermal amplification from both RNA or DNA targets in a single tube format. 

Benefits of SuperScript IV RT-LAMP Master Mix
• RNA and DNA target detection using Bst DNA polymerase for isothermal amplification in as little as five minutes
• Robust and specific cDNA synthesis using SuperScript IV Reverse Transcriptase
• Enhanced specificity and sensitivity with buffer formulation and RNaseOUT recombinant ribonuclease inhibitor
• Compatible with real-time and endpoint detection—includes SYTO 9 dye for real-time detection
• Flexible design provides several options for evaluating results, including real-time and endpoint detection methods

SuperScript IV RT-LAMP Master Mix includes an evolved Bst DNA polymerase mutant with a fast reaction speed and increased sensitivity and tolerance to inhibitors, as well as SuperScript IV Reverse Transcriptase with increased processivity, thermostability, and inhibitor resistance. To prevent RNA degradation by carry-over RNases, the master mix includes the potent, noncompetitive RNaseOUT Recombinant Ribonuclease Inhibitor. Our proprietary reaction buffer composition is optimized to increase specificity by delaying no-template control (NTC) amplification time.

Specifications

Product Type
Master Mix
For Use With (Application)
LAMP, RT-LAMP
Polymerase
Bst DNA Polymerase
Quantity
1 x 1.25 mL
No. of Reactions
100 Reactions
Optimal Reaction Temperature
65°C
Reverse Transcriptase
SuperScript IV
Shipping Condition
Dry Ice
Detection Method
Real-time and end-point detection
Includes Label or Dye
SYTO 9 Green Fluorescent Nucleic Acid Stain
Concentration
2X
Reaction Time
5 to 15 minutes
Unit Size
100 reactions

Contents & Storage

• 2X SuperScript IV RT-LAMP Master Mix (1.25 mL)
• SYTO Green Fluorescent Nucleic Acid Stain (30 μL)

Store at –20°C and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text