Informational:Informational:Save up to 31% on your order of cell culture products. View promotion details »
Gibco™

Fetal Bovine Serum, Value, One Shot™ format (formerly USDA-approved in North America or qualified, Brazil in other regions)

Catalog number: A5209401
Have Questions?
Gibco™

Fetal Bovine Serum, Value, One Shot™ format (formerly USDA-approved in North America or qualified, Brazil in other regions)

Catalog number: A5209401
Have Questions?
Catalog Number
A5209401
Unit Size
50 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
10 x 50 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
500 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A520940150 mL
-
A520940210 x 50 mL
-
A5256701500 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
  • Sera category: Value
  • Sera application: Cell culture with robust cell lines, good value for basic research
  • Testing: Up to 30 quality tests
  • Endotoxin level: ≤20 EU/mL (levels routinely ≤10 EU/mL)
  • Hemoglobin level: ≤25 mg/dL
  • Origin: Refer to CoA for specific origin

All products may not be available in all regions due to importation regulations

US Department of Agriculture (USDA) safety tested for the absence of bluetongue and akabane from countries that are USDA-approved for importation into the US

Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that help you meet your research needs and budget requirements.

Gibco Serum Delivers

  • ISO 13485-certified, processed in FDA-registered facilities
  • Triple filtered at 0.1 μm
  • Gibco bottle is easier to use in hood, reduces risk of contamination, and helps you perform cell culture more consistently
  • Peel-off sticker on label provides handy reference for lot number and expiration date recording in lab notebook

Specifications

Age
Fetal
Species
Cattle/Bovine
Country of Origin
Refer to CoA for specific origin
Format
One Shot
Product Type
Fetal Bovine Serum
Quantity
50 mL
Sterility
Sterile
Sterilization Method
Triple-filtered, 0.1 μm
Tested For
Up to 30 quality tests including 9CFR virus testing* endotoxin, performance (*Please note: If manufactured in the UK, Full 9CFR except for Rabies and BlueTongue Virus which is tested via EMA)
Endotoxin Concentration
≤20 EU/mL
Green Features
Less waste
Hemoglobin Concentration
≤25 mg/dL
Purity or Quality Grade
Value
Product Line
One Shot™
Shelf Life
4 Years
Shipping Condition
Dry Ice
USDA Safety Testing Included
USDA Safety Testing Performed for Mexican and Australian origins

Contents & Storage

Store at <-10°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text