Invitrogen™

Attune™ CytPix™ Violet Laser Upgrade Kit

Catalog number: A52446
Have Questions?
Invitrogen™

Attune™ CytPix™ Violet Laser Upgrade Kit

Catalog number: A52446
Have Questions?
Catalog Number
A52446
Unit Size
1 kit
Price (USD)
Catalog NumberUnit SizePrice (USD)
A524461 kitRequest A Quote
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Attune CytPix Violet Laser Upgrade Kit is designed to expand the 405-nm excitation capabilities of the Attune CytPix Flow Cytometer to utilize the array of labels, dyes, and applications available with a violet laser. This upgrade kit enables the detection of stem cell side populations using Vybrant DyeCycle violet, Pacific Blue, green, and orange dyes, as well as many Qdot nanocrystals and violet-excitable polymer dyes.

Expanded capabilities
A great way to stretch your research dollars is through upgrades, thereby expanding the capabilities of your instrument while minimizing expenditure on new instruments. Through the upgrade path, you can configure your Attune CytPix Flow Cytometer with different laser configurations to meet your ever-changing research needs.

Simple installation
The Attune CytPix flow cytometer was designed to be upgradable and minimize the time it takes for a qualified service engineer to add additional capabilities. This red laser upgrade kit comes with all of the optical parts needed to upgrade the instrument in typically less than four hours.

Specifications

Color
Violet
Description
Laser upgrade
Excitation/Emission
407 nm
For Use With (Application)
Flow Cytometry
For Use With (Equipment)
Attune CytPix Flow Cytometer
Format
Kit
Includes
Laser upgrade
Product Line
DJM
Quantity
1 Ea.
Regulatory Status
RUO
Shipping Condition
Room Temperature

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text