Informational:Inquire about OEM or Commercial Supply version of this product here.
Applied Biosystems™

MagMAX™ Wastewater Ultra Nucleic Acid Isolation Kit

Catalog number: A52606
Have Questions?
Applied Biosystems™

MagMAX™ Wastewater Ultra Nucleic Acid Isolation Kit

Catalog number: A52606
Have Questions?
Catalog Number
A52606
Unit Size
100 preps
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A52606100 preps
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The MagMAX Wastewater Ultra Nucleic Acid Isolation Kit enables efficient, high-throughput nucleic acid extraction from wastewater samples. The kit utilizes MagMAX magnetic-bead technology, ensuring reproducible recovery of high-quality nucleic acid compatible with a broad range of applications, including real-time PCR and next-generation sequencing.

Features of the MagMAX Wastewater Ultra Nucleic Acid Isolation Kit include:
• Automation-ready protocols designed for KingFisher Flex, KingFisher APEX, and KingFisher Duo systems
• Fast procedure allows for 24–96 samples to be processed in ∼45 minutes
• Compatible with wastewater and sewage concentrated via ultracentrifugation, precipitation, filtration, or bead-based viral enrichment and non-concentrated samples
• Includes methods for 200 μL to 500 mL starting volumes of wastewater
• Elution volumes ranging from 50 to 100 μL

Wastewater and sewage surveillance is a useful tool for monitoring the regional spread of viral and bacterial pathogens. Surveillance at different levels of operation, such as a university dormitory or an entire city, has been shown to be a reliable way to detect the SARS-CoV-2 virus before it becomes widespread in a population. Wastewater surveillance can be used to monitor other viral targets such as influenza and norovirus. However, wastewater and sewage samples are some of the most challenging when it comes to extraction of RNA and DNA, until now.

Maximize sample input and nucleic acid recovery in minimal time
Up to 500 mL of wastewater can be processed, with virus concentration protocols available for filtration.

Compatible with non-concentrated, precipitated, and ultracentrifuged samples
The MagMAX Wastewater Ultra Nucleic Acid Isolation Kit is also compatible with samples that have been concentrated by ultracentrifugation or precipitation or with non-concentrated samples (up to 1 mL). An optimized protocol enables extraction of 200-μL samples in just 45 minutes for a 96-well plate on the KingFisher Flex or APEX system. 1-mL samples can be extracted in just 45 minutes for a 24-well plate. Sludge samples can also be extracted using this protocol. When the protocol is completed, pure RNA and DNA is concentrated in 50–100 μL of elution buffer.

If an automated, bead-based virus enrichment is desired, the MagMAX Wastewater Ultra Nucleic Acid Isolation Kit with Virus Enrichment is available, which includes Dynabeads for wastewater virus enrichment.

Benefits of MagMAX magnetic bead–based purification
Magnetic beads offer many benefits compared to other technologies for isolating nucleic acid. Beads bind the DNA and RNA more efficiently than glass-fiber filters, resulting in higher and more consistent yields. Additionally, because filters and vacuum manifolds are not used, there is no risk of filter clogging due to cellular particulates in samples.

The Dynabeads MyOne Silane contained in the MagMAX Wastewater Ultra Nucleic Acid Isolation Kit have an optimized silica-like surface chemistry with high specific surface area that allows efficient kinetics and high sensitivity in nucleic acid capture. These aspects make Dynabeads MyOne Silane ideal for the recovery of RNA and DNA. Once captured, these nucleic acids can then be eluted in of 50–100 μL of elution buffer for use in downstream applications.

Specifications

Isolation Technology
Magnetic Bead
Sample Type
Sludge, Wastewater
Elution Volume
50 to 200 μL
Final Product Type
RNA and DNA
For Use With (Equipment)
KingFisher™ Duo Prime, KingFisher™ Flex, KingFisher™ Apex
For Use With (Application)
RT-PCR, qPCR, cDNA library construction, NGS, microarray analysis, blot hybridization, Northern/Southern blotting, in vitro translation, nuclease protection assays, nucleic acid labeling, hybridization
High-throughput Compatibility
High-throughput Compatible
Quantity
100 Preps
Shipping Condition
Room Temperature
Test Time
Up to 29 min. when using automated protocol
Starting Material Amount
Sludge: ≤200 mg
Wastewater: ≤500 mL

Contents & Storage

• 80 mL Lysis Buffer; room temperature
• 50 mL Binding Solution; room temperature
• 200 mL Wash Buffer; room temperature
• 25 mL Elution Solution; room temperature
• 4 mL Proteinase K; room temperature
• 2 mL DNA/RNA Binding Beads; room temperature

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text