The QuantStudio Absolute Q Digital PCR Starter Kit is designed to confirm the performance of the QuantStudio Absolute Q DigitalRead more
Have Questions?
Catalog Number
Quantity
A52732
1 Kit
Catalog number A52732
Price (USD)
884.00
Each
-
Add to cart
Quantity:
1 Kit
Price (USD)
884.00
Each
Add to cart
Ask our AI about this Product
The QuantStudio Absolute Q Digital PCR Starter Kit is designed to confirm the performance of the QuantStudio Absolute Q Digital PCR System either at the time of system installation or as part of future, user-defined system verification procedures. The Starter Kit may also be used for training users on the QuantStudio Absolute Q Digital PCR System workflow.
The Starter Kit contains Absolute Q DNA Digital PCR Master Mix (5X) (Cat. No. A52490), Absolute Q dPCR Starter Assay, and CEPH DNA Control, providing a complete set of starter reagents and assay for use with the QuantStudio Absolute Q MAP16 Plate Kit (Cat. No. A52865).
For Research Use Only. Not for use in diagnostics procedures.
Specifications
For Use With (Equipment)QuantStudio™ Absolute Q™ Digital PCR System
FormatTube
Product LineAbsolute Q™
Quantity1 Kit
Shipping ConditionDry Ice
TechniqueDigital PCR
PCR MethodDigital PCR
Unit SizeEach
Contents & Storage
1 - Absolute Q™ DNA Digital PCR Master Mix 1- Absolute Q™ dPCR Starter Assay (20 x conc) 1- Control DNA CEPH 1347-02. Store at -15°C to -25°C.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our
Privacy Notice.
Frequently asked questions (FAQs)
I finished running my experiment on the QuantStudio Absolute Q Digital PCR instrument but just realized that I had selected the wrong dyes during setup. Can I change the dyes that were used after the run?
If dye channels are turned off before the run, then data will not be collected for them as the run progresses. This means that changing the dyes after the run will not be possible.
I ran my experiment using the QuantStudio Absolute Q Digital PCR System but did not actually use dyes in all the channels. Is there a way to only see those dye channels that were used?
Unused dye channels can be turned off after the run by going to the SETUP tab and clicking EDIT SETUP and then EDIT GROUPS. Here, the Analysis can be changed to “Not Used.” After clicking SAVE twice, the data will be reanalyzed and the unused dye channels will not be present in the analysis.
I'm using the QuantStudio Absolute Q Digital PCR System and the software is saying that the concentration is NaN. What does that mean?
NaN stands for “not a number”. The software will display NaN instead of an actual number when it has detected some issue during the analysis of the array images.
With the QuantStudio Absolute Q Digital PCR instrument, the run data didn't automatically transfer after the run completed. How can I get the data?
We would recommend restarting the software. If the run still doesn't transfer, we would recommend closing the software, rebooting the instrument, and then starting the software. If further assistance is needed, please contact Technical Support at techsupport@thermofisher.com.
With the QuantStudio Absolute Q Digital PCR System, how much sample can be loaded into each well?
9 µL of sample is loaded into each well along with 15 µL of isolation buffer. Loading more or less of either the sample or isolation buffer will affect filling of the microchambers and the run results.