eBioscience™ Human Lysed Whole Blood Cell Flow Cytometry Workflow Kit
Product Image
Invitrogen™
eBioscience™ Human Lysed Whole Blood Cell Flow Cytometry Workflow Kit
The eBioscience Human Lysed Whole Blood Cell Flow Cytometry Workflow Kit contains all the buffers, blockers, and beads to completeRead more
Have Questions?
Catalog NumberQuantity
A530131 Kit
Catalog number A53013
Price (USD)
504.00
Each
-
Add to cart
Quantity:
1 Kit
Price (USD)
504.00
Each
Add to cart
Ask our AI about this Product
The eBioscience Human Lysed Whole Blood Cell Flow Cytometry Workflow Kit contains all the buffers, blockers, and beads to complete a standard flow cytometry experiment for staining cell surface targets on human lysed whole blood.
See protocol ›
This kit contains:
• UltraComp eBeads Plus Compensation Beads (Cat. No. 01-3333-41)
• eBioscience 10X RBC Lysis Buffer (multi-species) (00-4300-54)
• Fc Receptor Binding Inhibitor Polyclonal Antibody, eBioscience (14-9161-71)
• CellBlox Blocking Buffer (B001T06F01)
• eBioscience Flow Cytometry Staining Buffer (00-4222-57)
• eBioscience IC Fixation Buffer (00-8222-49)
When combined with the antibodies specific to your experiment, the reagents within this kit have been optimized to work together to detect cell surface targets by flow cytometry and broadly include:
• Effective compensation controls
• Fc receptor blocking
• Non-antibody, non-protein-based blocking solution for reducing background staining from fluorophores
• Antibody binding buffer that minimizes non-specific binding of antibodies
• Fixation buffer for storage of stained samples
Specifications
Quantity1 Kit
For Use With (Application)Flow Cytometry
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link