Thermo Scientific™

Pierce™ High Capacity Protein A MagBeads, alkali stable

Catalog number: A53038
Have Questions?
Thermo Scientific™

Pierce™ High Capacity Protein A MagBeads, alkali stable

Catalog number: A53038
Have Questions?
Catalog Number
A53038
Unit Size
Each
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
Each
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
Each
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
Each
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A53038Each
-
A53035Each
-
A53036Each
-
A53037Each
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Pierce High-Capacity Protein A MagBeads, Alkali Stable, are high-performance magnetic beads for purification of antibodies from cell culture supernatant, ascites, serum, and crude extracts.

Features of the High-Capacity Protein A MagBeads include:
• High binding capacity—≥ 40 mg HIgG/mL settled beads
• Alkaline stable—engineered to withstand standard CIP procedures with 0.1 M NaOH, enabling efficient cleaning of the beads for reuse
• Animal component-free raw materials
• Versatile—beads are compatible with manual and automated workflows (e.g., KingFisher instruments)
• Stable—high-affinity Protein A is securely immobilized to magnetic agarose to help prevent leaching of ligand, target antibody, or base support

The protein A ligand has been genetically modified for stability in the presence of NaOH, so the beads can be regenerated at least 10 times without sacrificing binding capacity. The ligand is produced in yeast and is immobilized on magnetic agarose with an average particle size of ∼30 μm.

Pierce High-Capacity Protein A MagBeads, Alkali-Stable, are suitable for purifying mAbs and a subpopulation of scFv’s and Fab fragments containing a VH3 domain. The beads are compatible with automation, allowing for higher throughput purifications and screening of purification conditions.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Binding Capacity
≥40 mg HIgG/mL settled beads
Description
High Capacity Magnetic Agarose Beads
Particle Size
∼ 30 μm
Quantity
4 x 25 mL
For Use With (Application)
Antibody Purification
Ligand Type
Alkaline Stable Protein A
Format
Bottle

Contents & Storage

Provided in the form of a 25% slurry.

Upon receipt store at 2–8°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text