Smart Deep™ Basecaller, 1-year license
Product Image
Applied Biosystems™
Smart Deep™ Basecaller, 1-year license
Smart Deep Basecaller is an improved basecaller for use with Sequencing Analysis Software 8. This license enables use of SmartRead more
Have Questions?
Catalog NumberQuantity
A532121 license
Catalog number A53212
Price (USD)
6,020.00
Each
Add to cart
Quantity:
1 license
Recurring order eligible. Learn more »
Price (USD)
6,020.00
Each
Add to cart
Ask our AI about this Product
Smart Deep Basecaller is an improved basecaller for use with Sequencing Analysis Software 8. This license enables use of Smart Deep Basecaller for 1 year.
Relative to KB Basecaller (included with Sequencing Analysis Software 8), this improved basecaller provides:
• Increased read lengths—more high quality basecalls at 5’ and 3’ ends
• More accurate pure and mixed base calls
• Improved basecalling accuracy through artifacts such as dye blobs, N-1 peaks, and mobility shifts
• Improved basecalling accuracy through heterozygous indel variants
• Reduced manual review time—fewer edits and fewer false positives
• Optional enhanced view trace visualization with a cleaned-up baseline and increased resolution in the 3’ end of plasmid (pure base) sequences
Note: SDB basecalling is enabled for data from SeqStudio Flex, SeqStudio, 3730, and 3500 series instruments.
The Smart Deep Basecaller license key is delivered by email and is then entered in Sequencing Analysis Software 8 to activate the license.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Data EntryCE Sequencing (.ab1 files)
License1 Year
Operating SystemWindows 10
Product TypeSmart Deep Basecaller 1 Year License
Quantity1 license
Software TypeSequencing Data Analysis
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Share catalog number, name or link
1x1 image pixel for data collection