Gibco™

Efficient-Pro™ Medium

Catalog number: A5322201
Have Questions?
Gibco™

Efficient-Pro™ Medium

Catalog number: A5322201
Have Questions?
Catalog Number
A5322201
Unit Size
1000 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
10 L
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
20 L
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A53222011000 mL
-
A532220210 L
-
A532220320 L
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Efficient-Pro Medium is a next-generation Chinese hamster ovary (CHO) medium used in the production of monoclonal antibodies (mAbs), offering significant performance and ease-of-use improvements over earlier Gibco and competitor products. Designed to work optimally with Efficient-Pro Feed 1 and Efficient-Pro Feed 2 for use in CHO-K1 and DG-44/CHO-S cell lines respectively, the Efficient-Pro system increases titer and specific productivity, improves protein quality and metabolite profiles, and is easier to use with less solubility issues than other CHO media and feed offerings.

The Efficient-Pro system will help you:
• Sustain cell viability for longer bioreactor runs and maximized titer
• Improve downstream outputs such as protein glycosylation, charge-variants, and aggregation
• Speed up process development and streamline transfer to manufacturing scale
• Increase flexibility with a range of format and packaging configurations
• Decrease cell shock with simplified adaptation protocols

Efficient-Pro Medium has been formulated as a chemically defined, animal-origin-free cell culture medium used in the bioproduction of mAbs and other therapeutics in traditional batch and fed-batch bioprocessing workflows, particularly in combination with CHO cell line variants.

Efficient-Pro Medium is a balanced formulation designed to meet the metabolic needs of mAb production and does not contain growth factors or insulin to ensure batch-to-batch consistency and production efficiency. Demonstrated to improve productivity in a CHO-K1 cell line when paired with Efficient-Pro Feed 1 compared to an industry-leader’s media and feed system, the Efficient-Pro system helps accelerate your time-to-market and achieve success.

Specifications

Cell Line
CHO-DG44, CHO-S, CHO-K1, GS-CHO
Classification
Animal Origin-free, Chemically-defined, Protein-free, Serum-free
Culture Type
Mammalian Cell Culture
Form
Liquid
Green Features
Sustainable packaging
Quantity
1000 mL
Shipping Condition
Room temperature

Contents & Storage

Store in the dark at 2–8°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text