Gibco™

Efficient-Pro™ AGT™ Medium

Catalog number: A5322304
Have Questions?
Gibco™

Efficient-Pro™ AGT™ Medium

Catalog number: A5322304
Have Questions?
Catalog Number
A5322304
Unit Size
500 L
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1 L
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
10 L
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
100 L
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A5322304500 L
-
A53223011 L
-
A532230210 L
-
A5322303100 L
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Efficient-Pro AGT Medium is a next-generation Chinese hamster ovary (CHO) medium used in the production of monoclonal antibodies (mAbs), offering significant performance and ease-of-use improvements over earlier Gibco and competitor products. Designed to work optimally with Efficient-Pro AGT Feed 1 and Efficient-Pro AGT Feed 2 for use in CHO-K1 and DG-44/CHO-S cell lines respectively, the Efficient-Pro AGT system increases titer and specific productivity, improves protein quality and metabolite profiles, and is easier to use with less solubility issues than other CHO media and feed offerings.

Efficient-Pro AGT system will help you:
• Benefit from the ease-of-use associated with Gibco’s trademarked, patented Advanced Granulation technology (AGT) for auto-pH and simplified dissolution
• Sustain cell viability for longer bioreactor runs and maximized titer
• Improve downstream outputs such as protein glycosylation, charge-variants, and aggregation
• Speed up process development and streamline transfer to manufacturing scale
• Increase flexibility with a range of format and packaging configurations
• Decrease cell shock with simplified adaptation protocols

Efficient-Pro AGT Medium has been formulated as a chemically defined, animal-origin-free cell culture medium used in the bioproduction of mAbs and other therapeutics in traditional batch and fed-batch bioprocessing workflows, particularly in combination with CHO cell line variants. The proprietary AGT format is the only one of its kind in the industry and is the most advanced media format available in the market.

Efficient-Pro AGT Medium is a balanced formulation designed to meet the metabolic needs of mAb production and does not contain growth factors or insulin to ensure batch-to-batch consistency and production efficiency. Demonstrated to improve productivity in a CHO-K1 cell line when paired with Efficient-Pro AGT Feed 1 compared to an industry-leader’s media and feed system, the Efficient-Pro AGT system helps accelerate your time-to-market and achieve success.

Specifications

Cell Line
CHO-DG44, CHO-S, CHO-K1, GS-CHO
Classification
Animal Origin-free, Chemically-defined, Protein-free, Serum-free
Culture Type
Mammalian Cell Culture
Form
Powder (AGT)
Green Features
Sustainable packaging
Quantity
500 L
Shipping Condition
Room temperature

Contents & Storage

Store in the dark at 2–8°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text