SAE Administrator Console Software v2.1 provides security, audit, and electronic signature (SAE) for the Applied Biosystems SeqStudio genetic analyzers. TheRead more
Have Questions?
Catalog Number
Quantity
A53717
1 license
Catalog number A53717
Price (EUR)
-
Quantity:
1 license
SAE Administrator Console Software v2.1 provides security, audit, and electronic signature (SAE) for the Applied Biosystems SeqStudio genetic analyzers. The software supports compliance with guidelines on electronic records security and prevents unauthorized access to the instrument.
Features of this software include: • System security–controls user access to the software. A default Administrator useraccount is provided, and additional user accounts and permissions can be user-defined. • Auditing–tracks actions performed by users and changes to the SAE module settings.The software automatically audits some actions silently. You can select other items forauditing and specify the audit mode. The auditing function provides reports for auditedSAE module changes and actions. • Electronic signature (e-signature)–determines if users are required to provide a username and password when performing certain functions. You can configure e-signature sothat a user can print a report or start a run only if the associated data are signed. You canalso configure each e-signature event to require multiple signatures and to require userswith specific permissions to sign.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)Sequencing Instruments (Capillary Electrophoresis-Based)