Genexus™ Barcodes 1-48 AS Kit
Genexus™ Barcodes 1-48 AS Kit
Ion Torrent™

Genexus™ Barcodes 1-48 AS Kit

The Ion Torrent Genexus Barcodes 1-48 AS Kit provides one plate filled with 48 dried-down, single-use barcodes and covered withRead more
Have Questions?
Catalog number A54129
Price (USD)/ 48 reactions
480.00
-
Add to cart
Price (USD)/ 48 reactions
480.00
Add to cart
Genexus™ Barcodes 1-48 AS Kit
Catalog numberA54129
Price (USD)/ 48 reactions
480.00
-
Add to cart

The Ion Torrent Genexus Barcodes 1-48 AS Kit provides one plate filled with 48 dried-down, single-use barcodes and covered with a pierceable foil seal, reducing the risk of cross contamination between wells. The barcodes can be used in increments of 1 to 48, until all barcodes are exhausted. When used in conjunction with Genexus Library Strips 1 and 2-AS, this kit supports flexible, fully automated preparation and normalization of up to 48 Ion AmpliSeq libraries. See also the Genexus Barcodes 49-96 AS Kit.

As part of the Genexus Integrated Sequencer workflow, the Genexus Barcodes 1-48 AS Kit:

  • Automates library preparation for 200–400 base-read Ion AmpliSeq libraries
  • Supports intake variability of clinical research samples in a cost-effective manner
  • Maximizes flexibility with the ability prepare 1 to 48 single-pool libraries per run
  • Enables parallel processing of up to four compatible assays in a single run
  • Delivers fast and simple instrument setup, with less than five minutes of hands-on time

The Genexus Integrated Sequencer automates library preparation, sequencing, analysis, and reporting in a hands-off, walkaway workflow that can deliver results in a single day. The innovative, multilane sequencing chip is designed to enable cost-effective processing of 1 to 48 single pool libraries.

For Research Use Only.
Specifications
FormatPre-filled Reagent Plates
Sample TypeDNA and RNA
Sequencing TypeGenome, DNA, and RNA Sequencing
For Use With (Equipment)Genexus™ Integrated Sequencer
For Use With (Application)Sequencing
LibrariesTargeted Sequencing Library
No. of Reactions48 Reactions
Product LineGenexus™
Product TypeBarcode Plate
Shipping ConditionRoom Temperature
Workflow StepLibrary Generation
Unit Size48 reactions