CTS™ AAV-MAX Transfection Kit
CTS™ AAV-MAX Transfection Kit
Gibco™

CTS™ AAV-MAX Transfection Kit

The Gibco CTS AAV-MAX Transfection Kit is an essential component of the CTS AAV-MAX Helper-Free AAV Production System. It isRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A54277011 Kit (For 1 L of production)
A54277021 Kit (For 10 L of AAV production)
A54277031 Kit (For 100 L of production)
Catalog number A5427701
Price (USD)
2,120.65
Online Exclusive
2,315.00
Save 194.35 (8%)
Each
-
Add to cart
Quantity:
1 Kit (For 1 L of production)
Price (USD)
2,120.65
Online Exclusive
2,315.00
Save 194.35 (8%)
Each
Add to cart
Ask our AI about this Product

The Gibco CTS AAV-MAX Transfection Kit is an essential component of the CTS AAV-MAX Helper-Free AAV Production System. It is used for the transient transfection of CTS Viral Production Cells 2.0. The kit includes a cationic lipid-based transfection reagent, as well as a transfection booster and production enhancer, which are designed to work together to produce adeno-associated virus (AAV) particles from CTS Viral Production Cells 2.0 when cultured in CTS Viral Production Medium.

Features of the CTS AAV-MAX Transfection Kit include:

  • Components work synergistically within the CTS AAV-MAX system to produce high AAV titers greater than 5 x 1010 vg/mL (unpurified)
  • Robust transfection protocols adaptable to large scale transfection
  • Manufactured in conformity with cGMP for medical devices (21 CFR Part 820) and following USP <1043> and European Pharmacopoeia (Ph. Eur.) 5.2.12 recommendations. Drug Master File and Regulatory Support File available upon request

Components of the kit include:

  • CTS AAV-MAX Transfection Reagent — cationic lipid-based transfection reagent for helper virus-free triple transfection of high-density cultures
  • CTS AAV-MAX Booster — chemically defined component that works with Transfection Reagent to achieve high transfection efficiency
  • CTS AAV-MAX Enhancer — chemically defined component that enhances the production of AAV particles

The CTS AAV-MAX Helper-Free AAV Production System is a scalable, high-titer platform for suspension AAV production with research-grade (Cat. No. A51217) and GMP versions to enable seamless transition from research to commercial manufacturing. Other components of the CTS AAV-MAX system include CTS Viral Production Cells 2.0 (A48400), CTS Viral Production Medium (A5144001), CTS Viral-Plex Complexation Buffer (A5145401), and Viral Production Medium, AGT (A5147101).

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.
Specifications
Expression SystemMammalian
Packaging TypeBottle
Phenol Red IndicatorNo
Product LineAAV-MAX
Product TypeTransfection kit for AAV production
Quantity1 Kit (For 1 L of production)
Shelf Life12 Months
FormatKit
Transfection TechniqueLipid-based Transfection, Transient Transfection
Unit SizeEach
Contents & Storage
• 1 x 6 mL bottle of CTS AAV-MAX Transfection Reagent
• 1 x 3 mL bottle of CTS AAV-MAX Transfection Booster
• 1 x 10 mL bottle of CTS AAV-MAX Enhancer

Store at 2°C to 8°C. Do not freeze.

Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link